Skip to Content
Merck
All Photos(1)

Key Documents

538493

Sigma-Aldrich

cis-2-Hexene

95%

Synonym(s):

(2Z)-2-Hexene, (Z)-2-Hexene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH3
CAS Number:
Molecular Weight:
84.16
Beilstein:
1719019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.396 (lit.)

bp

68-70 °C (lit.)

density

0.669 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC\C=C/C

InChI

1S/C6H12/c1-3-5-6-4-2/h3,5H,4,6H2,1-2H3/b5-3-

InChI key

RYPKRALMXUUNKS-HYXAFXHYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-2-Hexene is one of the isomeric forms of hexene. It is obtained from 2-hexyne, via semihydrogenation in the presence of Pd78(phen) [phen = 1,10-Phenanthroline] on titanium dioxide in n-octane. cis-2-Hexene is formed as one of the primary product from the dimerization of propylene in the presence of a nickel oxide-silica-alumina catalyst. It undergoes isomerization/hydrosilylation reaction with phenyl silane in the presence of cobalt catalyst to afford 1-hexylphenylsilane.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid, Regioconvergent, Solvent-Free Alkene Hydrosilylation with a Cobalt Catalyst.
Chen C, et al.
Journal of the American Chemical Society, 137(41), 13244-13247 (2015)
The dimerization of propylene over a nickel oxide-silica-alumina catalyst.
Imai H, et al.
Bulletin of the Chemical Society of Japan, 41(1), 45-48 (1968)
Synthesis and catalytic properties of large ligand stabilized palladium clusters.
Schmid G, et al.
J. Mol. Catal. A: Chem., 107(1), 95-104 (1996)
Chiara Giorio et al.
Faraday discussions, 200, 559-578 (2017-06-06)
Ozonolysis of alkenes is a key reaction in the atmosphere, playing an important role in determining the oxidising capacity of the atmosphere and acting as a source of compounds that can contribute to local photochemical "smog". The reaction products of
Sören Reinhard et al.
ChemMedChem, 12(17), 1464-1470 (2017-07-18)
Cationic lipo-oligomers containing unsaturated oleic acid are potent siRNA carriers based on electrostatic and hydrophobic lipo-polyplex formation and endosomal membrane destabilization. Lipo-oligomers can be produced by solid-phase-supported synthesis in sequence-defined form. However, the trifluoroacetic acid (TFA)-mediated removal of acid-labile protecting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service