Skip to Content
Merck
All Photos(1)

Key Documents

320323

Sigma-Aldrich

1-Hexene

97%

Synonym(s):

1-n-Hexene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1209240
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

vapor pressure

155 mmHg ( 21.1 °C)

Assay

97%

form

liquid

autoignition temp.

487 °F

refractive index

n20/D 1.388 (lit.)

bp

60-66 °C (lit.)

density

0.678 g/mL at 25 °C (lit.)

SMILES string

CCCCC=C

InChI

1S/C6H12/c1-3-5-6-4-2/h3H,1,4-6H2,2H3

InChI key

LIKMAJRDDDTEIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Hexene can be used:
  • As a substrate for polymerization by various catalysts.
  • To prepare good quality middle distillates and lubricating oils by oligomerization.
  • In the synthesis of aldehydes using supercritical CO2 and rhodium catalyst by hydroformylation reaction.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oligomerization of 1-hexene and 1-octene over solid acid catalysts.
de Klerk A.
Industrial & Engineering Chemistry Research, 44(11), 3887-3893 (2005)
Hydroformylation of 1-hexene in supercritical carbon dioxide using a heterogeneous rhodium catalyst. 3. Evaluation of solvent effects.
Hemminger O, et al.
Green Chemistry, 4(5), 507-512 (2002)
Stereospecific living Ziegler- Natta polymerization of 1-hexene.
Jayaratne, Kumudini C and Sita, Lawrence R
Journal of the American Chemical Society, 122(5), 958-959 (2000)
Isospecific living polymerization of 1-hexene by a readily available nonmetallocene C 2-symmetrical zirconium catalyst.
Tshuva, Edit Y et al.
Journal of the American Chemical Society, 122(43), 10706-10707 (2000)
Chia-Hsiu Chen et al.
Journal of the American Chemical Society, 137(21), 6965-6971 (2015-05-12)
The stereochemistry, kinetics, and mechanism of olefin polymerization catalyzed by a set of zirconium-based metallocenes was studied by NMR using dissolution dynamic nuclear polarization (DNP). Hyperpolarized 1-hexene was polymerized in situ with a C2 symmetric catalyst, [(EBI)ZrMe][B(C6F5)4] (EBI = rac-(C2H4(1-indenyl)2))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service