Skip to Content
Merck
All Photos(1)

Documents

110892

Sigma-Aldrich

trans-2-Hexene

97%

Synonym(s):

(E)-2-Hexene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH3
CAS Number:
Molecular Weight:
84.16
Beilstein:
1719020
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1 (vs air)

Quality Level

vapor pressure

263 mmHg ( 37.7 °C)

Assay

97%

autoignition temp.

473 °F

refractive index

n20/D 1.393 (lit.)

bp

68-69 °C (lit.)

mp

−98.5-−98 °C (lit.)

density

0.669 g/mL at 25 °C (lit.)

SMILES string

CCC\C=C\C

InChI

1S/C6H12/c1-3-5-6-4-2/h3,5H,4,6H2,1-2H3/b5-3+

InChI key

RYPKRALMXUUNKS-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-2-hexene is formed by the double bond shift and cis-trans isomerization which can be studied using microreactor-GLC techniques.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-13.0 °F - closed cup

Flash Point(C)

-25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mechanism of double-bond isomerization of olefins on solid acids.
Brouwer DM et al.
J. Catal., 1(1), 22-31 (1962)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service