Skip to Content
Merck
All Photos(2)

Key Documents

483311

Sigma-Aldrich

Bismuth(III) phosphate

99.99%

Synonym(s):

Bismuth orthophosphate, Phosphoric acid bismuth(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiO4P
CAS Number:
Molecular Weight:
303.95
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.99%

reaction suitability

core: bismuth
reagent type: catalyst

mp

>350 °C (lit.)

density

6.32 g/mL at 25 °C (lit.)

SMILES string

[Bi+3].[O-]P([O-])([O-])=O

InChI

1S/Bi.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+3;/p-3

InChI key

SFOQXWSZZPWNCL-UHFFFAOYSA-K

Application

Used for:
  • Coprecipitation of polonium
  • Approach to bismuth modifying procedure for voltammetric thick film carbon containing electrodes
  • Investigations of the association between plutonium radiation exposure and leukemia mortality
  • Membrane electrodes selective for hydrogen phosphate ions

Catalyst for:
  • Ammoxidation of propylene
  • Decomposition of CCl2F2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Holmberg et al.
Scandinavian journal of urology and nephrology, 23(3), 207-212 (1989-01-01)
One hundred and fifty-six patients with simple renal cysts had either a percutaneous puncture alone, percutaneous puncture combined with bismuth-phosphate instillation or no intervention at all. At follow-up, 25% of the cysts in which there was no intervention had grown
Chengsi Pan et al.
Environmental science & technology, 44(14), 5570-5574 (2010-06-29)
A high photocatalytic BiPO(4) with a novel nonmetal oxy acid structure is synthesized by a hydrothermal method. BiPO(4) photocatalyst has an optical indirect band gap of 3.85 eV. In a comparison of BiPO(4) with that of TiO(2) (P25, Degussa), it
M Dimberg et al.
International urology and nephrology, 20(2), 155-157 (1988-01-01)
Thirty-eight testicular hydroceles were treated by injection with 0.2-0.4 g of bismuth phosphate. The follow-up was completed in 32 cases, 17 (53%) of which showed complete regression although one patient had to wait 24 months for full regression. Eight patients
Minglei Zhao et al.
Inorganic chemistry, 52(2), 807-815 (2012-12-28)
Metal phosphates have been popularly regarded as excellent luminescence hosts of lanthanide ions, while such an issue is challenged by the ignorance about the structural stability that may originate from the different chemical nature between the framework and the dopant
Gregg J Lumetta et al.
Environmental science & technology, 43(20), 7843-7848 (2009-11-20)
A sample of high-level radioactive tank waste was characterized to provide a basis for developing a waste simulant. The simulant is required for pilot-scale testing of pretreatment processes in a nonradiological facility. The waste material examined was derived from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service