Skip to Content
Merck

244635

Tin(II) sulfate

≥95%

Synonym(s):

Stannous sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
SnSO4
CAS Number:
Molecular Weight:
214.77
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12161600
EC Number:
231-302-2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95%

reaction suitability

core: tin, reagent type: catalyst

SMILES string

[SnH2++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.Sn/c1-5(2,3)4;/h(H2,1,2,3,4);/q;+2/p-2

InChI key

OBBXFSIWZVFYJR-UHFFFAOYSA-L

Application

General applications:
  • Tin(II) sulfate can be used in the electrosynthesis of Cu2ZnSnS4 (CZTS) thin films for solar cell application.
  • It can be used to synthesize ternary Cu-Sn-S chalcogenides such as Cu2SnS3, Cu5Sn2S7 and Cu3SnS4 by successive ionic layer adsorption and reaction (SILAR) method.
  • SnSO4 can also be used in the preparation of tin (IV) oxide (SnO2) nanoparticles which are widely used in battery electrodes, fabrication of supercapacitors and opto-electronic devices.



Still not finding the right product?

Explore all of our products under Tin(II) sulfate


signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

target_organs

Cardio-vascular system, Respiratory system

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges

Storage Class

11 - Combustible Solids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


[Effect of tin sulfate on various indicators of lipid metabolism in pregnant albino rats].
N V Grin' et al.
Gigiena i sanitariia, (3)(3), 94-95 (1989-03-01)
Fabrication of ternary Cu?Sn?S sulfides by a modified successive ionic layer adsorption and reaction (SILAR) method.
Su Z, et al.
Journal of Materials Chemistry, 22(32), 16346-16352 (2012)
Surfactant-free solution phase synthesis of monodispersed SnO 2 hierarchical nanostructures and gas sensing properties.
Zhang H, et al.
CrystEngComm, 14(9), 3169-3176 (2012)



Global Trade Item Number

SKUGTIN
244635-100G04061825760162
244635-500G04061825760179