All Photos(2)
About This Item
Linear Formula:
(CH3O)2C6H3CH2Br
CAS Number:
Molecular Weight:
231.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
mp
69-70 °C (lit.)
functional group
bromo
storage temp.
2-8°C
SMILES string
COc1cc(CBr)cc(OC)c1
InChI
1S/C9H11BrO2/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5H,6H2,1-2H3
InChI key
BTHIGJGJAPYFSJ-UHFFFAOYSA-N
Related Categories
General description
3,5-Dimethoxybenzyl bromide is an aromatic bromide.
Application
3,5-Dimethoxybenzyl bromide may be used in the synthesis of:
- 3,4,3′,5′-tetramethoxystilbene
- 3,5-dimethoxyphenylacetic acid
- 1-(3,5-dimethoxyphenyl)-4-pentene
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthese de Δ8 et Δ9-tetrahydrocannabinol deuteries et trities.
Hoellinger H, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 13(3), 401-415 (1977)
An efficient synthesis of 3,5-dimethoxyhomophthalic acid, a key intermediate for synthesis of natural isocoumarins.
Saeed A, et al.
Journal of Heterocyclic Chemistry, 40(3), 519-522 (2003)
Dorota Chełminiak-Dudkiewicz et al.
Journal of photochemistry and photobiology. B, Biology, 181, 1-13 (2018-02-27)
Three magnesium sulfanyl porphyrazines differing in the size of peripheral substituents (3,5-dimethoxybenzylsulfanyl, (3,5-dimethoxybenzyloxy)benzylsulfanyl, 3,5-bis[(3,5-bis[(3,5-dimethoxybenzyloxy)benzyloxy]benzylsulfanyl) were exposed to visible and ultraviolet radiation (UV A + B + C) in order to determine their photochemical properties. The course of photochemical reactions in dimethylformamide solutions and the
Synthesis of resveratrol, DMU-212 and analogues through a novel Wittig-type olefination promoted by nickel nanoparticles.
Alonso F, et al.
Tetrahedron Letters, 50(25), 3070-3073 (2009)
Synthesis of pinosylvin and related heartwood stilbenes.
Bachelor FW, et al.
Canadian Journal of Chemistry, 48(10), 1554-1557 (1970)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service