Skip to Content
Merck
All Photos(1)

Documents

B17905

Sigma-Aldrich

Benzyl bromide

reagent grade, 98%

Synonym(s):

α-Bromotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Br
CAS Number:
Molecular Weight:
171.03
Beilstein:
385801
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Quality Level

vapor density

5.8 (vs air)

Assay

98%

form

liquid

impurities

≤0.5% p-bromotoluene

refractive index

n20/D 1.575 (lit.)

bp

198-199 °C (lit.)

mp

−3-−1 °C (lit.)

density

1.438 g/mL at 25 °C (lit.)

SMILES string

BrCc1ccccc1

InChI

1S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

AGEZXYOZHKGVCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl bromide is an aromatic halide that is mainly used as a protecting agent for protecting hydroxy group of alcohols via O-benzylation reaction.

Application

Benzyl bromide has been used as an initiator in the synthesis of poly(styrene-b-methylmethacrylate) copolymers by atom transfer radical polymerization. It can undergo Menschutkin reaction with 1, 2-dimethylimidazole to form 3-benzyl-1, 2-dimethylimidazolium bromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanderlisa Rita Meleti et al.
Acta tropica, 202, 105248-105248 (2019-11-05)
This paper reports the synthesis of (±)-licarin A 1, a dihydrobenzofuran neolignan, resultant of an oxidative coupling reaction of isoeugenol and horseradish peroxidase (HRP) enzyme. Following, three semi-synthetic derivatives from this compound were obtained: benzylated (±)-licarin A 2, methylated (±)-licarin
Giuseppe Belletti et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 26(4), 839-844 (2019-10-31)
Viedma ripening is a deracemization process that has been used to deracemize a range of chiral molecules. The method has two major requirements: the compound needs to crystallize as a conglomerate and it needs to be racemizable under the crystallization
Rate constants for reaction of 1, 2-dimethylimidazole with benzyl bromide in ionic liquids and organic solvents.
Skrzypczak A and Neta P.
International Journal of Chemical Kinetics, 36(4), 253-258 (2004)
Hon Man Yau et al.
Molecules (Basel, Switzerland), 14(7), 2521-2534 (2009-07-28)
While there has been much effort in recent years to characterise ionic liquids in terms of parameters that are well described for molecular solvents, using these to explain reaction outcomes remains problematic. Herein we propose that many reaction outcomes in
Virender Kumar et al.
Theranostics, 8(15), 4033-4049 (2018-08-22)
Rationale: Successful treatment of pancreatic cancer remains a challenge due to desmoplasia and prevalence of KRAS mutation. While hedgehog (Hh) ligand levels are upregulated in pancreatic cancer cells and contribute to desmoplasia, there is significant downregulation of tumor suppressor let-7b

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service