Skip to Content
Merck
All Photos(2)

Documents

209384

Sigma-Aldrich

3-Methoxybenzyl chloride

97%

Synonym(s):

3-(Chloromethyl)anisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2Cl
CAS Number:
Molecular Weight:
156.61
Beilstein:
636684
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

contains

potassium carbonate as stabilizer

refractive index

n20/D 1.544 (lit.)

bp

124 °C/13 mmHg (lit.)

density

1.078 g/mL at 25 °C (lit.)
1.078 g/mL

storage temp.

2-8°C

SMILES string

COc1cccc(CCl)c1

InChI

1S/C8H9ClO/c1-10-8-4-2-3-7(5-8)6-9/h2-5H,6H2,1H3

InChI key

VGISFWWEOGVMED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methoxybenzyl chloride was used to alkylate 8-benzyloxy-2(1H)-quinolinone in the presence of DMF and NaH. It was also used in the synthesis of diarylmethanes via in situ organozinc-mediated, palladium-catalyzed cross-coupling between benzyl and aryl halide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

N-vs O-Alkylation in 2 (1H)-quinolinone derivatives.
Guo Z-X, et al.
Tetrahedron Letters, 40(38), 6999-7002 (1999)
Prachi Singh et al.
SLAS discovery : advancing life sciences R & D, 22(4), 440-446 (2017-03-23)
Analysis of interactions between molecules is of fundamental importance in life science research. In this study, we applied weak affinity chromatography, based on high-performance liquid chromatography and mass spectrometry, as a powerful tool for direct analysis of the components of
Christophe Duplais et al.
Chemical communications (Cambridge, England), 46(4), 562-564 (2010-01-12)
A remarkably simple entry to unsymmetrical diarylmethanes has been developed that relies on an in situ organozinc-mediated, palladium-catalyzed cross-coupling. Thus, by mixing a benzyl and aryl halide together in the presence of Zn metal and a Pd catalyst, diarylmethanes are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service