479861
2,4-Hexadiene, mixture of isomers
technical grade, 90%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH=CHCH=CHCH3
CAS Number:
Molecular Weight:
82.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical grade
Assay
90%
impurities
10% positional double bond isomers
refractive index
n20/D 1.445 (lit.)
bp
82 °C (lit.)
density
0.72 g/mL at 25 °C (lit.)
SMILES string
C\C=C\C=C\C
InChI
1S/C6H10/c1-3-5-6-4-2/h3-6H,1-2H3/b5-3+,6-4+
InChI key
APPOKADJQUIAHP-GGWOSOGESA-N
Looking for similar products? Visit Product Comparison Guide
General description
2,4-Hexadiene, mixture of isomers, is a conjugated diene. The interaction between palladium chloride and 2,4-hexadiene has been reported to proceed in a stereospecific manner. The photodissociation spectra of 2,4-hexadiene isomers have been recorded. The polymers obtained by the polymerization of 2,4-hexadiene isomers are reported to have the trans-1,4 structure.
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
7.0 °F - closed cup
Flash Point(C)
-13.9 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Photodissociation spectroscopy of hexadiene radical ions: structural rearrangements in the gas-phase ions.
Benz RC, et al.
Journal of the American Chemical Society, 103(7), 1799-1802 (1981)
The stereospecific addition of dienes to palladium chloride.
Lukas J, et al.
Journal of Organometallic Chemistry, 47(1), 153-163 (1973)
The Polymerization of 2,4-Hexadiene. III. Cationic Polymerizability of 2,4-Hexadiene Isomers.
Kamachi M, et al.
Polymer Journal, 1(5), 499-504 (1970)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service