Skip to Content
Merck
All Photos(1)

Key Documents

145491

Sigma-Aldrich

2,3-Dimethyl-1,3-butadiene

98%, contains 100 ppm BHT as stabilizer

Synonym(s):

2,3-Dimethylbuta-1,2-diene, 2,3-Dimethylenebutane, Biisopropenyl, Diisopropenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)C(CH3)=CH2
CAS Number:
Molecular Weight:
82.14
Beilstein:
605285
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

269 mmHg ( 37.7 °C)

Quality Level

Assay

98%

form

liquid

contains

100 ppm BHT as stabilizer

refractive index

n20/D 1.438 (lit.)

bp

68-69 °C (lit.)

mp

−76 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)C(C)=C

InChI

1S/C6H10/c1-5(2)6(3)4/h1,3H2,2,4H3

InChI key

SDJHPPZKZZWAKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Dimethyl-1,3-butadiene (DMBD) is a conjugated diene. It undergoes Diels Alder cycloaddition reaction with 2-thio-3-chloroacrylamides under thermal, catalytic and microwave conditions. It undergoes thermal [4+2] cycloaddition reaction with 3-acetyl-, 3-carbamoyl and 3-ethoxycarbonylcoumarins under solvent free conditions. DMBD participates in polymerization reactions in the presence of iron dichloride complexes based catalysts.

Application

2,3-Dimethyl-1,3-butadiene was used to investigate the reactions of 1,3-dienes with the Si(001) surface using scanning tunneling microscopy and fourier transform infrared spectroscopy.
It may be used in the following processes:
  • Preparation of 1,3,6-triene derivatives of corresponding 1-aryl-substituted 1,3-dienes by 1,4-hydrobutadienylation in the presence of cobalt catalyst.
  • Synthesis of 6-aryl(hetaryl)-3,4-dimethyl-1-nitro-1-cyano-3-cyclohexenes by reacting with gem-cyanonitroethenes.
  • As a halogen trap during the study of the photolysis reaction of dibromo adduct of 2,5-diphenyltellurophene.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxy radical kinetics resulting from the OH-initiated oxidation of 1,3-butadiene, 2,3-dimethyl-1,3-butadiene and isoprene.
Jenkin ME, et al.
Journal of Atmospheric Chemistry, 29(3), 267-298 (1998)
Polymerization of 1, 3-dienes with iron complexes based catalysts: Influence of the ligand on catalyst activity and stereospecificity.
Ricci G, et al.
J. Mol. Catal. A: Chem., 204, 287-293 (2003)
Cobalt-Catalyzed 1, 4-Hydrobutadienylation of 1-Aryl-1,3-dienes with 2,3-Dimethyl-1,3-butadiene.
Bohn MA, et al.
Angewandte Chemie (International Edition in English), 50(41), 9689-9693 (2011)
Marie Kissane et al.
Organic & biomolecular chemistry, 8(24), 5602-5613 (2010-10-12)
The Diels-Alder cycloadditions of cyclopentadiene and 2,3-dimethyl-1,3-butadiene to a range of 2-thio-3-chloroacrylamides under thermal, catalytic and microwave conditions is described. The influence of reaction conditions on the outcome of the cycloadditions, in particular the stereoselectivity and reaction efficiency, is discussed.
Aryl (hetaryl)-containing gem-cyanonitroethenes: Synthesis, structure, and reactions with 2,3-dimethyl-1,3-butadiene.
Baichurin RI, et al.
Russ. J. Gen. Chem., 85(8), 1845-1854 (2015)

Articles

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

The Diels–Alder reaction is the reaction between a conjugated diene and an alkene (dienophile) to form unsaturated six-membered rings. It is also referred to as a cycloaddition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service