Skip to Content
Merck
All Photos(1)

Key Documents

259470

Sigma-Aldrich

Allyl ether

98%

Synonym(s):

Diallyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCH2)2O
CAS Number:
Molecular Weight:
98.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

94-95 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

C=CCOCC=C

InChI

1S/C6H10O/c1-3-5-7-6-4-2/h3-4H,1-2,5-6H2

InChI key

ATVJXMYDOSMEPO-UHFFFAOYSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daham Jeong et al.
Carbohydrate polymers, 198, 563-574 (2018-08-11)
A novel self-healing hydrogel was prepared from a cationic β-cyclodextrin oligomer allyl ether [C(βCD-OM)AE] using a triple cross-linking strategy combining electrostatic interaction, host-guest complexation, and CC bonds as the macrocrosslinker. Here, the C(βCD-OM)AE@Ad gel was successfully prepared by polymerization of
Weike Zou et al.
Science advances, 6(13), eaaz2362-eaaz2362 (2020-04-08)
Dynamic covalent polymer networks exhibit unusual adaptability while maintaining the robustness of conventional covalent networks. Typically, their network topology is statistically nonchangeable, and their material properties are therefore nonprogrammable. By introducing topological heterogeneity, we demonstrate a concept of topology isomerizable
Xuemei Wang et al.
Talanta, 166, 300-305 (2017-02-19)
Novel magnetic mesoporous molecularly imprinted polymers (MMIPs) with core-shell structure were prepared by simple surface molecular imprinting polymerization using tetrabromobisphenol-S (TBBPS) as the template. The MMIPs-TBBPS were characterized by fourier-transform infrared spectrometry (FTIR), scanning electron microscopy (SEM), transmission electron microscopy
Wen Fu et al.
Journal of the mechanical behavior of biomedical materials, 95, 53-59 (2019-04-08)
It has already been reported that incorporation of thiol-ene into methacrylate resin could reduce shrinkage stress significantly. However, the mass fraction of thiol-ene in thiol-ene-methacrylate resin system should be limited because of deterioration of mechanical properties. In order to increase
Mohammad A Rahman et al.
Toxicology letters, 292, 31-38 (2018-04-26)
Diallyl sulfide (DAS) has been shown to prevent xenobiotic (e.g. ethanol, acetaminophen) induced toxicity and disease (e.g. HIV-1) pathogenesis. DAS imparts its beneficial effect by inhibiting CYP2E1-mediated metabolism of xenobiotics, especially at high concentration. However, DAS also causes toxicity at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service