Skip to Content
Merck
All Photos(1)

Key Documents

450944

Sigma-Aldrich

Iron(II) chloride

AnhydroBeads, −10 mesh, 99.9% trace metals basis

Synonym(s):

Ferrous chloride, Iron dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2
CAS Number:
Molecular Weight:
126.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis

product line

AnhydroBeads

Quality Level

Assay

99.9% trace metals basis

reaction suitability

reagent type: catalyst
core: iron

impurities

≤1500.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

677 °C (lit.)

density

3.16 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[Fe]Cl

InChI

1S/2ClH.Fe/h2*1H;/q;;+2/p-2

InChI key

NMCUIPGRVMDVDB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of a novel cis-Fe(BPE5)2Cl2 (BPE5=1,2-diphospholanoethane) complex, which has potential application in many types of reactions such as intra- or intermolecular activation.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Field, L.D. et al.
Inorganic Chemistry, 37, 612-612 (1998)
Qinqin Xia et al.
The Journal of organic chemistry, 77(20), 9366-9373 (2012-10-03)
Iron-catalyzed direct C-N bond formation between azoles and amides is described. The oxidative coupling reactions of sp(3) C-H bonds adjacent to a nitrogen atom in amides and sulfonamides with the N-H bond in azoles proceeded smoothly in the presence of
Jiangfei Meng et al.
Journal of food science, 77(1), C8-14 (2011-12-21)
Wines made from 3 spine grape (Vitis davidii Foex) genotypes-Junzi 1# (JZ 1#), Junzi 2# (JZ 2#), and Liantang (LT)-and Cherokee rose (Rosa laevigata Michx., CR) were evaluated for their phenolics composition and antioxidant activities by several assays, including 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulfonic
Sofia Tresintsi et al.
Water research, 46(16), 5255-5267 (2012-07-25)
Various iron oxy-hydroxides were synthesized in a continuous flow kilogram-scale production reactor through the precipitation of FeSO(4) and FeCl(2) in the pH range 3-12 under intense oxidative conditions to serve as arsenic adsorbents. The selection of the optimum adsorbent and
Shaoru Chen et al.
PloS one, 8(1), e53662-e53662 (2013-01-24)
This study was designed to investigate the potentially protective effects of glycyrrhetinic acid (GA) and the role of transcription factor nuclear factor-erythroid 2(NF-E2)-related factor 2 (Nrf2) signaling in the regulation of Carbon Tetrachloride (CCl(4))-induced chronic liver fibrosis in mice. The

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service