Skip to Content
Merck
All Photos(7)

Key Documents

157740

Sigma-Aldrich

Iron(III) chloride

greener alternative

reagent grade, 97%

Synonym(s):

Ferric chloride, Iron trichloride, Molysite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
powder

grade

reagent grade

Quality Level

vapor density

5.61 (vs air)

vapor pressure

1 mmHg ( 194 °C)

Assay

97%

form

powder

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

304 °C (lit.)

greener alternative category

SMILES string

Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) chloride is a mild oxidizing agent and participates in various carbon-carbon-bond forming reactions. On crystallization with water, it forms hydrates. It can be prepared by reacting chlorine with the scrap iron at approximately 650°C.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Iron(III) chloride (Ferric Chloride, FeCl3) has been used as a surface modulator for the characterization of wines by fingerprinting technology. It has been used for the quantitative estimation of sulfide in various biological samples.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resitance properties.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Iron (III) chloride-based mild synthesis of phenanthrene and its application to total synthesis of phenanthroindolizidine alkaloids"
Wang L-K, et al.
Tetrahedron, 64(32), 7504-7510 (2008)
Novel non-specific liquid fingerprint technology for wine analysis: a feasibility study.
Siivonen JJ, et al.
Australian Journal of Grape and Wine Research, 20(2), 172-177 (2014)
"Iron (III) chloride-catalyzed convenient one-pot synthesis of homoallyl benzyl ethers starting from aldehydes"
Watahiki T, et al.
Organic Letters, 5(17), 3045-3048 (2003)
Yuanyuan Kong et al.
Cell death & disease, 10(9), 624-624 (2019-08-20)
Recent findings demonstrate that aberrant downregulation of the iron-exporter protein, ferroportin (FPN1), is associated with poor prognosis and osteoclast differentiation in multiple myeloma (MM). Here, we show that FPN1 was downregulated in MM and that clustered regularly interspaced short palindromic
H F Hazlett et al.
Scientific reports, 10(1), 10935-10935 (2020-07-04)
Cystic fibrosis (CF) is a genetic disease caused by mutations in the CF transmembrane conductance regulator (CFTR) gene, resulting in chronic bacterial lung infections and tissue damage. CF macrophages exhibit reduced bacterial killing and increased inflammatory signaling. Iron is elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service