Skip to Content
Merck
All Photos(3)

Key Documents

448303

Sigma-Aldrich

16-Mercaptohexadecanoic acid

90%

Synonym(s):

MHDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)15CO2H
CAS Number:
Molecular Weight:
288.49
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

90%

form

solid

mp

65-69 °C

SMILES string

OC(=O)CCCCCCCCCCCCCCCS

InChI

1S/C16H32O2S/c17-16(18)14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-19/h19H,1-15H2,(H,17,18)

InChI key

INOAASCWQMFJQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

16-Mercaptohexadecanoic acid (MHA) is a long chained alkanethiol which forms self-assembled monolayers (SAMs) on a variety of surfaces.

Application

MHA forms SAMs on gold dipyramids which can be used in the fabrication of nanoresonators for shell-isolated nanoparticle enhanced raman spectroscopy (SHINERS). It can also be used to surface modify gold electrodes that can be used for sensitive detection of clozapine. Gold surfaces can be self-assembled with MHA by polymer pen lithography (PPL) which can be potentially used in bioengineering.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Landon J Brower et al.
Beilstein journal of nanotechnology, 8, 2307-2314 (2017-11-29)
Integration of surface-anchored metal-organic frameworks (surMOFs) within hierarchical architectures is necessary for potential sensing, electronic, optical, or separation applications. It is important to understand the fundamentals of film formation for these surMOFs in order to develop strategies for their incorporation
Modification of surfaces of silver nanoparticles for controlled deposition of silicon, manganese, and titanium dioxides.
Abdulrahman HB, et al.
Applied Surface Science, 427, 334-339 (2018)
Sensitive detection of clozapine using a gold electrode modified with 16-mercaptohexadecanoic acid self-assembled monolayer.
Huang Fei, et al.
Talanta, 72(2), 457-462 (2007)
Brandon H Bowser et al.
Nanomaterials (Basel, Switzerland), 8(9) (2018-08-26)
Metal-organic frameworks (MOFs) are extremely porous, crystalline materials with high surface area for potential use in gas storage, sequestration, and separations. Toward incorporation into structures for these applications, this study compares three variations of surface-bound and free-standing HKUST-1 MOF structures:
Andreas Mark et al.
Small (Weinheim an der Bergstrasse, Germany), 15(43), e1902976-e1902976 (2019-09-24)
The colloidal probe technique, which is based on the atomic force microscope, revolutionizes direct force measurements in many fields, such as interface science or biomechanics. It allows for the first time to determine interaction forces on the single particle or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service