Skip to Content
Merck
All Photos(1)

Documents

674508

Sigma-Aldrich

(11-Mercaptoundecyl)tetra(ethylene glycol)

90%

Synonym(s):

MUTEG, (1-Mercaptoundec-11-yl)tetra(ethylene glycol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)11(OCH2CH2)4OH
Molecular Weight:
380.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

90%

refractive index

n20/D 1.475

density

0.997 g/mL at 25 °C

storage temp.

−20°C

SMILES string

OCCOCCOCCOCCOCCCCCCCCCCCS

InChI

1S/C19H40O5S/c20-10-12-22-14-16-24-18-17-23-15-13-21-11-8-6-4-2-1-3-5-7-9-19-25/h20,25H,1-19H2

InChI key

IOBMVUDHPRVCSO-UHFFFAOYSA-N

General description

(11-Mercaptoundecyl)tetra(ethylene glycol) (MUTEG) is a capping agent that is used in the surface modification of a variety of nanoparticles. It can be also be used as an effective protein repellent.

Application

MUTEG can be used to functionalize gold nanoparticles, which can potentially be used to signal peptides, as fluorescent probes and as electrocatalytic membranes. It can also facilitate the sensing of protein on polymeric substrates for biosensor based applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic electrocatalytic effect of nanostructured mixed films formed by functionalised gold nanoparticles and bisphthalocyanines
Medina-Plaza C, et al.
Analytica Chimica Acta, 851(3), 95-102 (2014)
Plasmonic flow-through biosensor using a polymeric substrate.
Buchenauer A, et al.
Journal of Micromechanics and Microengineering, 24(3), 034001-034001 (2014)
Acrylate-Facilitated Cellular Uptake of Gold Nanoparticles
Krpetic V, et al.
Small, 7(14), 1982-1986 (2011)
Multiple biomarkers biosensor with just-in-time functionalization: application to prostate cancer detection
Parra-Cabrera C, et al.
Biosensors And Bioelectronics, 77(3), 1192-1200 (2016)

Articles

We presents an article regarding Self-Assembled Monolayers: Advantages of Pure Alkanethiols.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service