Skip to Content
Merck
All Photos(1)

Key Documents

439770

Sigma-Aldrich

Gadolinium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Gadolinium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤150.0 ppm Trace Rare Earth Analysis

SMILES string

Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd/h3*1H;/q;;;+3/p-3

InChI key

MEANOSLIBWSCIT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gadolinium(III) chloride is a white inorganic halide hygroscopic solid soluble in water. Gd3+ species are particularly intriguing because they possess the highest known number of unpaired spins among all elements. The exceptional magnetic properties and symmetry of Gd3+ make it a valuable component in both NMR spectroscopy and MRI applications. It is widely used as a dopant/precursor to enhance the optical and magnetic properties of various materials used in MRI contrast agents, electronic devices, and phosphors.

Application

Gadolinium(III) chloride can be used:
  • As a precursor to synthesize Gd metal nanoparticles.
  • As a dopant to prepare CdSe nanoparticles as a contrast agent for optical and magnetic resonance imaging applications.
  • To prepare Gd-Hematoporphyrin monomethyl ether(HMME) phosphors for oxygen sensing.
  • To fabricate radioluminescent silica optical fibers via surface plasma chemical vapor deposition (SPCVD), for magneto-optical devices.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparison study on the influence of the central metal ions in palladium (II)-and gadolinium (III)-porphyrins for phosphorescence-based oxygen sensing
Lixin Zang, et al.
Journal of Materials Chemistry, 4, 9581-9587 (2016)
Lillian Zhang et al.
The Journal of toxicological sciences, 37(2), 447-453 (2012-04-03)
Liver toxicity is a side effect observed with some herbal treatments, including Piper methysticum. The possible mechanisms responsible include inflammation subsequent to activation of liver macrophages and oxidative damage. Hepatotoxicity of the pharmacologically active component of Piper methysticum (kavalactones) was
Puja Gulati et al.
Naunyn-Schmiedeberg's archives of pharmacology, 386(3), 255-264 (2012-12-12)
The present study was designed to investigate the potential of gadolinium, a stretch-activated calcium channel blocker in ischemic reperfusion (I/R)-induced brain injury in mice. Bilateral carotid artery occlusion of 12 min followed by reperfusion for 24 h was given to induce cerebral
T Hasezaki et al.
Die Pharmazie, 66(9), 698-703 (2011-10-27)
Nanomaterials have potential toxicity that is not found in micromaterials, and it is therefore essential to understand their biological activity and potential toxicity. We focused on silica nanoparticles, since it was previously reported that the intravenous administration of silica nanoparticles
Chikara Abe et al.
Journal of applied physiology (Bethesda, Md. : 1985), 114(1), 28-36 (2012-11-10)
Water drinking is known to induce the pressor response. The efferent pathway in this response involves sympathoexcitation, because the pressor response was completely abolished by ganglionic blockade or an α(1)-adrenergic antagonist. However, the afferent pathway in this response has not

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service