Skip to Content
Merck
All Photos(2)

Key Documents

334065

Sigma-Aldrich

Indium(III) chloride

98%

Synonym(s):

Indium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
InCl3
CAS Number:
Molecular Weight:
221.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
98%
form:
powder or flakes (or chunks)
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder or flakes (or chunks)

reaction suitability

reagent type: catalyst
core: indium

density

3.46 g/mL at 25 °C (lit.)

SMILES string

Cl[In](Cl)Cl

InChI

1S/3ClH.In/h3*1H;/q;;;+3/p-3

InChI key

PSCMQHVBLHHWTO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III) chloride (InCl3) is an indium halide that is a Lewis acid in a variety of organic reactions.[1]

Application

InCl3 can be used for a variety of application such as:
  • fabrication of a photoanode for dye sensitized solar cells[2]
  • a catalyst in Michael addition of silylenol and indoles[1]

Useful catalyst for aqueous organic reactions including C-C bond formation, aldol reactions, and reductions.[3] An efficient, yet mild, Lewis acid for organic synthesis.[4]

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Babu, S.A.
Synlett, 531-531 (2002)
Satoko Fujimoto et al.
Bioscience, biotechnology, and biochemistry, 66(6), 1389-1392 (2002-08-07)
(R)-(-)-Muscone (3-methylcyclopentadecanone, 1) the key perfumery component isolated from the male musk deer, Moschus moschiferus,* was synthesized from the easily available chiral building block, (R)-3-tert-butoxycarbonyl-2-methylpropanoic acid (2), by employing ring-closing olefin metathesis (RCM). Antipode (+)-1 was also synthesized in a
Indium (III) chloride-catalyzed Michael addition of thiols to chalcones: a remarkable solvent effect
Ranu BC, et al.
ARKIVOC (Gainesville, FL, United States), 3, 44-50 (2005)
Fabrication and characterization of ZnO: In thin film as photoanode for DSSC using natural fruit dyes
AIP Conference Proceedings, 1660(1), 070049-070049 (2015)
Naveen Mulakayala et al.
Bioorganic & medicinal chemistry letters, 22(15), 5063-5066 (2012-07-04)
A convenient and practical methodology for the synthesis of 2-aryl quinazolin-4(3H)-ones by the condensation of o-aminobenzamides with aromatic aldehydes under mild conditions using catalytic InCl(3) with good yields and high selectivities. This method has been extended for the synthesis of

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service