Skip to Content
Merck
All Photos(2)

Key Documents

G7532

Sigma-Aldrich

Gadolinium(III) chloride hexahydrate

99% (titration)

Synonym(s):

Gadolinium trichloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3 · 6H2O
CAS Number:
Molecular Weight:
371.70
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99% (titration)
form:
powder
Pricing and availability is not currently available.

Quality Level

Assay

99% (titration)

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

density

2.42 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

PNYPSKHTTCTAMD-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Gadolinium(III) chloride hexahydrate (GdCl3.6H2O) can be used as a metal precursor in applications such as dual mode MRI contrasting agents. It can also be used as a catalyst in the preparation of one-pot synthesis of α-amino nitriles via one pot three component reaction.[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuqi Yang et al.
Nano letters, 19(1), 441-448 (2018-12-19)
Nano contrast agents (Nano CA) are nanomaterials used to increase contrast in the medical magnetic resonance imaging (MRI). However, the related relaxation mechanism of the Nano CA is not clear yet and little significant breakthrough in relaxivity enhancement has been
Lanthanum (III) nitrate hexahydrate or gadolinium (III) chloride hexahydrate catalyzed one-pot synthesis of alpha-amino nitriles
Narasimhulu M, et al.
J. Mol. Catal. A: Chem., 264(1-2), 288-292 (2007)
T 1-T 2 dual-modal MRI contrast agents based on superparamagnetic iron oxide nanoparticles with surface attached gadolinium complexes
Szpak A, et al.
Journal of Nanoparticle Research, 16(11), 2678-2678 (2014)
Neil Phillips et al.
Human gene therapy, 25(7), 651-661 (2014-03-19)
Treatment of type 1 diabetes with gene transfer-induced cellular reprogramming requires a pancreatic transcription factor such as Neurogenin-3 (Ngn3) and as of yet unknown component of the adenoviral particle. Despite intensive study, there are many unsolved processes related to the
Simone Rodrigues Silva et al.
Bioinorganic chemistry and applications, 2015, 942147-942147 (2015-09-09)
Complexes of gadolinium(III) with N-octanoyl-N-methylglucamine (L8) and N-decanoyl-N-methylglucamine (L10) with 1 : 2 stoichiometry were synthesized and characterized by elemental analysis, electrospray ionization-tandem mass spectrometry (ESI-MS), infrared (IR) spectroscopy, and molar conductivity measurements. The transverse (r 2) and longitudinal (r 1) relaxivity

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service