Skip to Content
Merck
All Photos(2)

Documents

434108

Sigma-Aldrich

Calix[6]arene

97%

Synonym(s):

Calix[6]arenehexol, p-H-Calix[6]arene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H36O6
CAS Number:
Molecular Weight:
636.73
Beilstein:
4345238
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

395 °C (lit.)

SMILES string

Oc1c2Cc3cccc(Cc4cccc(Cc5cccc(Cc6cccc(Cc7cccc(Cc1ccc2)c7O)c6O)c5O)c4O)c3O

InChI

1S/C42H36O6/c43-37-25-7-1-8-26(37)20-28-10-3-12-30(39(28)45)22-32-14-5-16-34(41(32)47)24-36-18-6-17-35(42(36)48)23-33-15-4-13-31(40(33)46)21-29-11-2-9-27(19-25)38(29)44/h1-18,43-48H,19-24H2

InChI key

JLSWUKWFQCVKCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa A Shehata et al.
Journal of pharmaceutical and biomedical analysis, 146, 179-187 (2017-09-08)
Acquisition of the dissolution profiles of more than single active ingredient in a multi-analyte pharmaceutical formulation is a mandatory manufacturing practice that is dominated by utilization of the off-line separation-based chromatographic methods. This contribution adopts a new "Double-Track" approach with
Hui Zhao et al.
Nanoscale, 9(12), 4272-4282 (2017-03-16)
Alternative cleavage and polyadenylation (APA) is involved in several important biological processes in animals, e.g. cell growth and development, and cancer progression. The increasing data show that cancer cells are inclined to produce mRNA isoforms with a shortened 3'UTR undergoing
Safaa M Riad et al.
Journal of AOAC International, 101(3), 723-731 (2017-09-10)
Although the ultimate goal of administering active pharmaceutical ingredients (APIs) is to save countless lives, the presence of impurities and/or degradation products in APIs or formulations may cause harmful physiological effects. Today, impurity profiling (i.e., the identity as well as
Said A Hassan et al.
Mikrochimica acta, 187(9), 491-491 (2020-08-09)
The major objective of this work was to develop a portable, disposable, cost-effective, and reliable POC solid-state electrochemical sensor based on potentiometric transduction to detect benzodiazepine abuse, mainly diazepam (DZP), in biological fluids. To achieve that, microfabricated Cu electrodes on
Mohamed K Abd El-Rahman et al.
Talanta, 221, 121409-121409 (2020-10-21)
Choline (Ch+), is a vitamin-like essential water-soluble organic micronutrient. The US-FDA requires that infant formula not made from cow's milk must be supplemented with Ch+. Direct determination of Ch+ in milk powders and infant formulas is a challenging task due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service