Skip to Content
Merck

C4642

α-Cyclodextrin

≥98%

Synonym(s):

alpha-Cyclodextrin, α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
UNSPSC Code:
12352005
NACRES:
NA.22
PubChem Substance ID:
EC Number:
233-007-4
Beilstein/REAXYS Number:
79627
MDL number:
Assay:
≥98%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

mp

>278 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

Application

α-Cyclodextrin can be used:
  • In fabricating reversible supramolecular assemblies.
  • As a modifier to enhance the solubility of oleic acid stabilized nanoparticles.
  • In the preparation of polyrotaxanes with ply(ethylene glycol).
  • In the synthesis of cationic star polymers with oligoethylenimine.



Still not finding the right product?

Explore all of our products under α-Cyclodextrin


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library