Skip to Content
Merck
All Photos(1)

Key Documents

86142

Sigma-Aldrich

4-Sulfocalix[4]arene sodium salt

≥95.0% (HPLC)

Synonym(s):

25,26,27,28-Tetrahydroxycalix[4]arene-5,11,17,23-tetrasulfonic acid sodium salt, Calix[4]arene-4-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H24-xO16S4Nax
CAS Number:
Beilstein:
5221495
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (HPLC)

form

crystals

impurities

≤3% methanol

mp

≥300 °C (lit.)

SMILES string

[Na].Oc1c2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc1cc(c2)S(O)(=O)=O)c5O)S(O)(=O)=O)c4O)S(O)(=O)=O)c3O)S(O)(=O)=O

InChI

1S/C28H24O16S4.Na/c29-25-13-1-14-6-22(46(36,37)38)8-16(26(14)30)3-18-10-24(48(42,43)44)12-20(28(18)32)4-19-11-23(47(39,40)41)9-17(27(19)31)2-15(25)7-21(5-13)45(33,34)35;/h5-12,29-32H,1-4H2,(H,33,34,35)(H,36,37,38)(H,39,40,41)(H,42,43,44);/q;+1/p-1

InChI key

GZPNIEYNWYONCE-UHFFFAOYSA-M

General description

4-Sulfocalix[4]arene is a cyclic oligomer, which is prepared by hydroxyalkylation of phenol and aldehyde. It belongs to the family of calixarenes with repeating phenolic units connected via methylene groups forming a macrocyclic molecule. It has an amphiphilic nature with different functional groups that make it useful in drug delivery systems. It shows potential applications in various fields such as analysis and separation, enzyme mimetic systems and self-assembly membranes.

Application

4-Sulfocalix[4]arene sodium salt is an amphiphilic calix[n]arene that can be used as a coupling agent for organic molecules and metal complexes because of its hydrophobic (aromatic ring) and hydrophilic (SO3-) environments.

Other Notes

Water-soluble calixarene; complexing agent, e.g. for lanthanides; Regioselective acid catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Yoshida et al.
Chemistry Letters (Jpn), 2105-2105 (1991)
Binding of Ru (NH3) 5pz2+ to 4-Sulfocalix [4] arene Sodium Salt. Effects of the Host- Guest Interaction on Electron Transfer Processes
Lopez-Cornejo P, et al.
The Journal of Physical Chemistry B, 113(38), 12721-12726 (2009)
Lateral assembly of oxidized graphene flakes into large-scale transparent conductive thin films with a three-dimensional surfactant 4-sulfocalix [4] arene
Sundramoorthy AK, et al.
Scientific reports, 5, 10716-10716 (2015)
M. Komiyama et al.
Chemistry Letters (Jpn), 937-937 (1991)
N. Sato et al.
Chemistry Letters (Jpn), 1261-1261 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service