Skip to Content
Merck

1.03296

Chloroform-D1

0.03 vol.% TMS, deuteration degree min. 99.8% for NMR spectroscopy (stabilized with silver) MagniSolv

Synonym(s):

Chloroform-D1, Deuterochloroform

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
CCl3D
CAS Number:
Molecular Weight:
120.38
UNSPSC Code:
12142201
EC Index Number:
212-742-4
NACRES:
NA.22
MDL number:
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

vapor pressure

211 hPa ( 20 °C)

form

liquid

potency

695 mg/kg LD50, oral (Rat), >3980 mg/kg LD50, skin (Rabbit)

technique(s)

NMR: suitable

impurities

≤0.01% Water (H<SUB>2</SUB>O) (NMR), 0.03% Tetramethylsilane (NMR)

bp

60 °C/1013 hPa

mp

-64.1 °C

solubility

4.6 g/L

density

1.50 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)[2H]

InChI

1S/CHCl3/c2-1(3)4/h1H/i1D

InChI key

HEDRZPFGACZZDS-MICDWDOJSA-N

General description

A wide range of MagniSolv deuterated solvents with extremely low residual water, excellent chemical purity, and the highest isotopic enrichment available can satisfy the most demanding requirements of researchers. Depending on application and sensitivity of the NMR spectrometer our offer solvents with deuteration degrees between 98% and 99.96%. In case of all the water soluble deuterated standard products, water content is specified according to both Karl Fischer and NMR.

Analysis Note

Deuteration degree: ≥ 99.8 %
Water (NMR, H₂O): ≤ 0.01 %
Tetramethylsilane (NMR-spectrum): 0.03%
Performance test (NMR-spectrum): passes test

Other Notes

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Legal Information

MAGNISOLV is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Chloroform-D1


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
103296002504022536041193
103296250004027269165727
103296010004022536041209
103296050004022536744179