Skip to Content
Merck
All Photos(3)

Key Documents

431508

Sigma-Aldrich

Tin(II) chloride dihydrate

≥99.99% trace metals basis

Synonym(s):

Stannous chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2 · 2H2O
CAS Number:
Molecular Weight:
225.65
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥99.99% trace metals basis

reaction suitability

core: tin
reagent type: catalyst

bp

652 °C (lit.)
652 °C

mp

37-38 °C (dec.) (lit.)

cation traces

Ca: ≤0.005%
Fe: ≤0.003%
K: ≤0.005%
Na: ≤0.01%
Pb: ≤0.01%

SMILES string

O.O.Cl[SnH2]Cl

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Reducing agent; carbonyl allylation; Lewis acid catalyst in C-C bond-forming reactions; catalyst withAgClO4 for the synthesis of α-glycosides; synthesis of alkenes, dienes, cis-vinyloxiranes, and allylic selenides; deoxygenation of 1,4-endoperoxides; protection of carboxylic acids as 1,3-dithianes;selective p-methoxybenzyl ether cleavage reagent; additive in hydroformylation and carbonylation reactions.

Features and Benefits

Meets A.C.S. reagent specifications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Currie et al.
Inorganic chemistry, 52(4), 1710-1721 (2012-06-09)
The anionic speciation of chlorostannate(II) ionic liquids, prepared by mixing 1-alkyl-3-methylimidazolium chloride and tin(II) chloride in various molar ratios, χ(SnCl2), was investigated in both solid and liquid states. The room temperature ionic liquids were investigated by (119)Sn NMR spectroscopy, X-ray
Qiu-Sha Tang et al.
International journal of nanomedicine, 6, 3077-3085 (2011-12-14)
The purpose of this study was to develop intraperitoneal hyperthermic therapy based on magnetic fluid hyperthermia, nanoparticle-wrapped cisplatin chemotherapy, and magnetic particles of albumin. In addition, to combine the multiple-killing effects of hyperthermal targeting therapy, chemotherapy, and radiotherapy, the albumin-nanoparticle
Rajendran Suresh et al.
The Journal of organic chemistry, 77(3), 1468-1476 (2012-01-11)
The synthesis of 2-(1H-pyrrolo[2,3-b]pyridin-3-yl)quinolines by a SnCl(2)-catalyzed multicomponent reaction has been described. The reaction proceeds chemo- and regioselectively in an atom-economic way, generating a library of 24 quinoline derivatives.
Caitian Gao et al.
Nanoscale, 4(11), 3475-3481 (2012-05-11)
A high-efficiency photoelectrode for dye-sensitized solar cells (DSSCs) should combine the advantageous features of fast electron transport, slow interfacial electron recombination and large specific surface area. However, these three requirements usually cannot be achieved simultaneously in the present state-of-the-art research.
Liyan Zheng et al.
Dalton transactions (Cambridge, England : 2003), 41(5), 1630-1634 (2011-12-08)
It was found that stannous chloride (SnCl(2)), as a popular inorganic reducing reagent, could obviously enhance the electrochemiluminescence (ECL) of tris(2,2'-bipyridyl) ruthenium(II) (Ru(bpy)(3)(2+)) in aqueous solution. Some factors affecting the ECL reactions between Ru(bpy)(3)(2+) and Sn(2+), including pH, concentrations of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service