Skip to Content
Merck
All Photos(1)

Key Documents

1.07815

Supelco

Tin(II) chloride dihydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Tin(II) chloride dihydrate, Tin dichloride, dihydrate, Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sn · 2H2O
CAS Number:
Molecular Weight:
225.65
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-868-0
NACRES:
NA.21
Assay:
98.0-103.0% (SnCl2.2H2O basis, iodometric)
grade:
ACS reagent
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

98.0-103.0% (SnCl2.2H2O basis, iodometric)

form

solid

potency

700 mg/kg LD50, oral (Rat)

impurities

≤0.05% Substances not precipitated with H2S (as sulphates)

pH

1-2 (20 °C, 100 g/L in H2O)

bp

623 °C/1013 hPa (anhydrous substance)

mp

38 °C (anhydrous substance)

density

2.71 g/cm3 at 20 °C

bulk density

1250 kg/m3

anion traces

sulfate (SO42-): ≤0.002%

cation traces

As: ≤0.0001%
Ca: ≤0.005%
Cu: ≤0.001%
Fe: ≤0.002%
K: ≤0.005%
NH4+: ≤0.002%
Na: ≤0.01%
Pb: ≤0.005%
heavy metals (as Pb): ≤0.005%

storage temp.

15-25°C

SMILES string

[Sn](Cl)Cl.O.O

InChI

1S/2ClH.2H2O.Sn/h2*1H;2*1H2;/q;;;;+2/p-2

InChI key

FWPIDFUJEMBDLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • The Effect of Precursor Concentration on the Particle Size, Crystal Size, and Optical Energy Gap of Ce(x)Sn(1-x)O(2) Nanofabrication: This study explores the role of precursor concentration, including Tin(II) chloride dihydrate, in the control of particle size and crystal size during the fabrication of cerium-doped tin oxide nanomaterials. These parameters critically influence the optical properties and energy gap of the produced nanomaterials, making the compound valuable in applications like sensors, catalysts, and energy devices (Al-Hada NM et al., 2021).

Linkage

Replaces: SX0885-1

Analysis Note

Assay (iodometric, SnCl₂·2H₂O): 98.0 - 103.0 %
Identity: passes test
Appearance of solution in hydrochloric acid: passes test
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.005 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.01 %
NH₄ (Ammonium): ≤ 0.002 %
Pb (Lead): ≤ 0.005 %
Substances not precipitated with H₂S (as sulphates): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service