Skip to Content
Merck
All Photos(1)

Key Documents

426229

Sigma-Aldrich

Formic acid-d2

95 wt. % in D2O, 98 atom % D

Synonym(s):

Perdeuterioformic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCO2D
CAS Number:
Molecular Weight:
48.04
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

99% (CP)

form

liquid

concentration

95 wt. % in D2O

technique(s)

NMR: suitable
bio NMR: suitable

refractive index

n20/D 1.369 (lit.)

bp

100.8 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.273 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]OC([2H])=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)/i1D/hD

InChI key

BDAGIHXWWSANSR-PFUFQJKNSA-N

Related Categories

General description

Formic acid-d2 is isotopically labeled formic acid that is used to catalyze isoprene to criegee intermediates. It is used to improve NMR profiling of amino metabolites in biofluids.

Application

Formic acid-d2 is used in the synthesis of N-monodeuteriomethyl-2-substituted piperidines.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Ottolina et al.
Biochimica et biophysica acta, 998(2), 173-178 (1989-10-05)
The stereospecifically labeled coenzymes [4R-2H]NADH, [4R-2H]NADPH and [4S-2H]NAD(P)H were synthesized enzymatically in high yield and high isotopic purity (greater than or equal to 95%) with 2HCOO2H/formate dehydrogenase, (CH3)2C2HOH/alchol dehydrogenase from Thermoanaerobium brockii and [1-2H]glucose/glucose dehydrogenase, respectively. This set of deuterated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service