Skip to Content
Merck
All Photos(1)

Key Documents

489441

Sigma-Aldrich

Formic acid-d

95 wt. % in D2O, 98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2D
CAS Number:
Molecular Weight:
47.03
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Assay

99% (CP)

concentration

95 wt. % in D2O

refractive index

n20/D 1.369

bp

100.8 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.246 g/mL at 25 °C

mass shift

M+1

SMILES string

[H]C(=O)O[2H]

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)/i/hD

InChI key

BDAGIHXWWSANSR-DYCDLGHISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service