Skip to Content
Merck
All Photos(2)

Documents

550574

Sigma-Aldrich

Methanol-OD

99 atom % D

Synonym(s):

mono-Deuteromethanol, Methan(ol-d), Methyl alcohol-OD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OD
CAS Number:
Molecular Weight:
33.05
Beilstein:
1730948
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Assay

≥99.00%

form

liquid

expl. lim.

5.5-36.5 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.327 (lit.)

bp

65.5 °C (lit.)

mp

-99 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC

InChI

1S/CH4O/c1-2/h2H,1H3/i2D

InChI key

OKKJLVBELUTLKV-VMNATFBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol-OD is a deuterated form of methanol. It is used as a solvent in NMR-based research and analyses.

Application

Methanol-OD can be used as a deuterated solvent to:
  • Perform APPI HDX MS (atmospheric pressure photoionization hydrogen/deuterium exchange mass spectrometry).
  • Synthesis 1,4-butanediol-D6 (1,4-BD-D6)from 1,4-butanediol-D8.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehydration of alcohols over alumina. VIII. Ether formation from the deuterated methanols CH3OH, CD3OH, CH3OD, and CD3OD.
Knozinger H, et al.
The Journal of Physical Chemistry, 72(8), 2770-2774 (1968)
High Resolution Fourier Transform Far Infrared Spectroscopy of CH3OD.
Mukhopadhyay I, et al.
Journal of Molecular Spectroscopy, 186(1), 15-21 (1997)
The Raman Spectra of the Methyl Alcohols, CH3OH, CH3OD, and CH2DOD.
Halford JO, et al.
J. Chem. Phys. , 5(12), 927-932 (1937)
Infrared intensities of liquids XXI: integrated absorption intensities of CH3OH, CH3OD, CD3OH and CD3OD and dipole moment derivatives of methanol.
Bertie JE and Zhang SL.
Journal of Molecular Structure, 413, 333-363 (1997)
Photoionization of CH3OH, CD3OH, and CH3OD: Dissociative ionization mechanisms and ionic structures.
Berkowitz J.
J. Chem. Phys. , 69(7), 3044-3054 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service