Skip to Content
Merck
All Photos(3)

Key Documents

404861

Sigma-Aldrich

Tetrakis(hydroxymethyl)phosphonium chloride solution

80% in H2O

Synonym(s):

THPC solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)4PCl
CAS Number:
Molecular Weight:
190.56
Beilstein:
3631669
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

80% in H2O

refractive index

n20/D 1.512

density

1.341 g/mL at 25 °C

SMILES string

[Cl-].OC[P+](CO)(CO)CO

InChI

1S/C4H12O4P.ClH/c5-1-9(2-6,3-7)4-8;/h5-8H,1-4H2;1H/q+1;/p-1

InChI key

AKXUUJCMWZFYMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetrakis(hydroxymethyl)phosphonium chloride solution (80% in H2O) has been used as a reducing agent and stabilizing ligand for the synthesis of gold nanoparticles (AuNPs) from gold(III) chloride trihydrate (HAuCl4.3H2O). It is a tetra-functional, amine-reactive, aqueous crosslinker that can be used for tuning the properties of protein-based hydrogels for 3D cell encapsulation applications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile preparation of water-soluble fluorescent gold nanoclusters for cellular imaging applications.
Shang L, et al.
Nanoscale, 3(5), 2009-2014 (2011)
Beyond gold: rediscovering tetrakis-(hydroxymethyl)-phosphonium chloride (THPC) as an effective agent for the synthesis of ultra-small noble metal nanoparticles and Pt-containing nanoalloys
Hueso J L, et al.
Royal Society of Chemistry Advances, 3(26), 10427-10433 (2013)
D J Ball et al.
Journal of photochemistry and photobiology. B, Biology, 45(1), 28-35 (1998-11-20)
Three substituted zinc (II) phthalocyanines (one anionic, one cationic and one hydrophobic) have been compared to two clinically used photosensitisers, 5,10,15,20-tetra (m-hydroxyphenyl) chlorin (mTHPC) and polyhaematoporphyrin (PHP), as potential agents for photodynamic therapy (PDT). Oxygen-consumption experiments, performed to follow the
Christian Bayreder et al.
Medical physics, 33(7), 2506-2518 (2006-08-11)
Magnetic resonance (MR)-based polymer gel dosimetry using normoxic polymer gels, represents a new dosimetric method specially suited for high-resolution three-dimensional dosimetric problems. The aim of this study was to investigate the dose response with regard to stability, accuracy, reproducibility, and
Tobias Kiesslich et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 9(5), 734-743 (2010-04-02)
Photodynamic therapy (PDT) using Photofrin and, recently, Foscan has gained broad acceptance for palliative treatment of non-resectable cholangiocarcinoma (CC). No information, however, is available whether the phenotype of CC tumour cells has an effect on the efficiency of the treatment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service