Skip to Content
Merck
All Photos(3)

Key Documents

403245

Sigma-Aldrich

(3-Carboxypropyl)trimethylammonium chloride

technical grade

Synonym(s):

γ-Butyrobetaine hydrochloride, Deoxycarnitine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H16NO2Cl
CAS Number:
Molecular Weight:
181.66
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

mp

220 °C (dec.) (lit.)

SMILES string

[Cl-].C[N+](C)(C)CCCC(O)=O

InChI

1S/C7H15NO2.ClH/c1-8(2,3)6-4-5-7(9)10;/h4-6H2,1-3H3;1H

InChI key

GNRKTORAJTTYIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Adamiak et al.
ACS nano, 11(10), 9877-9888 (2017-10-04)
Cellular uptake by macrophages and ensuing clearance by the mononuclear phagocyte system stands as a significant biological barrier for nanoparticle therapeutics. While there is a growing body of work investigating the design principles essential for imparting nanomaterials with long-circulating characteristics
Carnitine synthesis in scorbutic guinea pigs.
Nutrition reviews, 43(6), 185-187 (1985-06-01)
W J Thoma et al.
Biochimica et biophysica acta, 797(1), 136-139 (1984-01-24)
The effect of ascorbate deficiency on carnitine biosynthesis was investigated in young male guinea pigs. Liver and skeletal muscle carnitine levels were reduced in scorbutic animals. Heart and kidney concentrations remained unchanged. 14C-labeled 4-N-trimethylaminobutyrate was administered to control, pair-fed and
L M Henderson et al.
Federation proceedings, 41(12), 2843-2847 (1982-10-01)
The biosynthesis of carnitine proceeds from trimethyllysine (TML) by beta-hydroxylation by a liver or kidney mitochondrial enzyme, which requires oxygen, alpha-ketoglutarate, ferrous iron, and ascorbate. This dioxygenase is rapidly inactivated by preincubation with Fe2+, but not Fe3+. The evidence suggests
J S Serrano et al.
Methods and findings in experimental and clinical pharmacology, 5(4), 251-254 (1983-01-01)
The effects of GABA, its quaternary derivative N-trimethyl GABA and quinidine have been studied in a model of ventricular automaticity induced in the isolated right ventricle of the rat. GABA shows low anti-automatic activity. N-trimethyl GABA and quinidine were, however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service