Skip to Content
Merck
All Photos(4)

Key Documents

B3501

Sigma-Aldrich

Betaine hydrochloride

≥99%

Synonym(s):

(Carboxymethyl)trimethylammonium hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2COOH · Cl-
CAS Number:
Molecular Weight:
153.61
Beilstein:
3916181
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥99%

form

powder or crystals

mp

241-242 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

[Cl-].C[N+](C)(C)CC(O)=O

InChI

1S/C5H11NO2.ClH/c1-6(2,3)4-5(7)8;/h4H2,1-3H3;1H

InChI key

HOPSCVCBEOCPJZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Betaine (trimethyl glycine) is an osmoprotectant, which is highly effective than proline. It is formed in from choline. Betaine is usually present in grains, spinach, beets, broccoli, shellfish and marine algae.

Application

Betaine hydrochloride has been used as a component of fixative in HEPES or cacodylate buffer to improve preservation of the ultrastructural morphology of human spermatozoa.

Biochem/physiol Actions

Betaine hydrochloride can decrease hepatotoxicity, that is stimulated by ethanol.
End-product of oxidative metabolism of choline, betaine is a general methyl donor, in particular in a minor pathway of methionine biosynthesis. It is used to treat homocystinuria, which is a defect in the major pathway of methionine biosynthesis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved preservation of ultrastructural morphology in human spermatozoa using betaine in the primary fixative
Swan MA
International Journal of Andrology, 20(1), 45- 54 (1997)
Can Phytochemicals be Effective in Preventing Ethanol-Induced Hepatotoxicity in the Geriatric Population? An Evidence-Based Revisit
Foods and Dietary Supplements in the Prevention and Treatment of Disease in Older Adults, 163- 170 (2015)
Effect of drying medium on the viability of dried
Selmer OE, et al.
Immobilized Cells: Basics and Applications, 11, 229-229 (1996)
Caroline E Geisler et al.
Cell reports, 35(13), 109298-109298 (2021-07-01)
Hepatic lipid accumulation in obesity correlates with the severity of hyperinsulinemia and systemic insulin resistance. Obesity-induced hepatocellular lipid accumulation results in hepatocyte depolarization. We have established that hepatocyte depolarization depresses hepatic afferent vagal nerve firing, increases GABA release from liver
Matthew J Szucs et al.
mBio, 11(5) (2020-10-01)
Viruses have developed innovative strategies to exploit the cellular machinery and overcome the antiviral defenses of the host, often using specifically structured RNA elements. Examples are found in the Flavivirus genus (in the family Flaviviridae), where during flaviviral infection, pathogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service