Skip to Content
Merck
All Photos(1)

Key Documents

395064

Sigma-Aldrich

Ethylamine solution

2.0 M in methanol

Synonym(s):

Aminoethane, Monoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5NH2
CAS Number:
Molecular Weight:
45.08
Beilstein:
505933
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10.07 psi ( 20 °C)
19.16 psi ( 55 °C)

Quality Level

form

liquid

concentration

2.0 M in methanol

density

0.81 g/mL at 20 °C
0.783 g/mL at 25 °C

functional group

amine

storage temp.

2-8°C

SMILES string

CCN

InChI

1S/C2H7N/c1-2-3/h2-3H2,1H3

InChI key

QUSNBJAOOMFDIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylamine is an alkali. Ethylamine modified vermiculite (Ethyl-VER) was prepared, which was used to remove cesium from aqueous solution. The oxidation of ethylamine on platinum single crystal electrodes in an acidic medium has been studied by cyclic voltammetry (CV) and Fourier transform infrared reflection-absorption spectroscopy (FT-IRRAS).

Application

Ethylamine solution may be employed as reaction medium in the synthesis of large-scale flower-like CuS microspheres.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang Long et al.
Journal of colloid and interface science, 428, 295-301 (2014-06-10)
Ethylamine modified vermiculite (Ethyl-VER) with high specific surface area and excellent pore structure was prepared to remove cesium from aqueous solution. The physic-chemical properties of the pristine and modified vermiculite were analyzed by X-ray diffraction (XRD), Fourier-transform infrared (FTIR), specific
Self-assembly of CuS nanoflakes into flower-like microspheres: synthesis and characterization.
Shen X-P, et al.
Journal of Physics and Chemistry of Solids, 70(2), 422-427 (2009)
Oxidation of methylamine and ethylamine on Pt single crystal electrodes in acid medium.
Huerta F, et al.
Journal of Electroanalytical Chemistry, 469(2), 159-169 (1999)
Câline Christine et al.
Organic & biomolecular chemistry, 10(46), 9183-9190 (2012-10-23)
The synthesis of a phosphonated acyclic bifunctional chelate L* for the labeling of biomaterial is described. L* is based on a pyridine backbone, functionalized in ortho positions by aminomethyl-bis-methylphosphonic acids, and, in the para position, by a side chain containing
Yuying Shuai et al.
Journal of the science of food and agriculture, 90(15), 2563-2567 (2010-08-26)
Theanine, a unique amino acid found almost exclusively in tea plants, has various favourable physiological and pharmacological functions in humans. Gamma-glutamyltranspeptidase (GGT, EC 2.3.2.2) is considered to be the most effective enzyme for the production of theanine. In fact, GGT

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service