Skip to Content
Merck
All Photos(2)

Key Documents

388246

Sigma-Aldrich

Dimethylamine solution

2.0 M in methanol

Synonym(s):

N,N-Dimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.90 -2.20 M (NaOH, titration)
2.0 M in methanol

density

0.775 g/mL at 25 °C

functional group

amine

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethylamine, a secondary amine, is used as a raw material for the production of many agrichemicals and pharmaceuticals. The solvents dimethylformamide (DMF) and dimethylacetamide (DMA) are derived from dimethylamine. It is also used as a precursor for the preparation of amido derivatives.

Application

Dimethylamine can be used as a precursor for the preparation of a number of chemical building blocks such as dimethyl(3-chloropropyl)amine , 2-chloro-N,N-dimethylethanimidamide , and 4, 6-dichloro-2-(dimethylamino)-1,3,5-triazine .

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Eyes, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

4-Dimethylaminobutyraldehyde diethyl acetal
FENG R-l, et al.
JOURNAL-SHANDONG UNIVERSITY OF SCIENCE AND TECHNOLOGY NATURAL SCIENCE, 5 (2011)
Synthesis of 2-amino-4-imino-4, 5-dihydrothiazoles from N-sulfonyl-α -chloroamidines and thiourea
Abdelaal SM and Bauer L
Journal of Heterocyclic Chemistry, 25(6), 1849-1856 (1988)
Methylamines
van Gysel A B and Musin W
Ullmann's Encyclopedia of Industrial Chemistry (1997)
Synthesis of 3-{4-[4-dimethylamino-6-(4-methyl-2-oxo-2H-chromen-7-yloxy)-[1, 3, 5] triazin-2-ylamino]-phenyl}-2-phenyl-5-(4-pyridin-2-yl-piperazin-1-ylmethyl)-thiazolidin-4-one and their biological evaluation
Patel D, et al.
Medicinal Chemistry Research, 21(10), 2926-2944 (2012)
L Lee et al.
Cancer research, 41(10), 3992-3994 (1981-10-01)
Using a method for nitrosamine analysis that gives high recovery values and that is free from artifactual synthesis of nitrosamines, we have shown that human feces do not contain volatile nitrosamines (detection limit, 0.1 to 0.5 microgram/kg). We also showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service