Skip to Content
Merck
All Photos(1)

Key Documents

391638

Sigma-Aldrich

Dimethylphosphoramidous dichloride

97%

Synonym(s):

(Dichlorophosphanyl)dimethylamine, (Dimethylamino)dichlorophosphine, Dichloro(dimethylamido)phosphorus, Dichloro(dimethylamino)phosphine, Dichloro-N,N-dimethylaminophosphine, N,N-Dimethylphosphoramidous dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NPCl2
CAS Number:
Molecular Weight:
145.96
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.505 (lit.)

bp

150 °C (lit.)

density

1.271 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CN(C)P(Cl)Cl

InChI

1S/C2H6Cl2NP/c1-5(2)6(3)4/h1-2H3

InChI key

XPWWDZRSNFSLRQ-UHFFFAOYSA-N

General description

Dimethylphosphoramidous dichloride (dimethylaminodichlorophosphine) is a phosphorus chloride derivative. It exists in two stable isomers, a gauche-conformer with C1 symmetry and anti-conformer with Cs symmetry. Its synthesis has been reported. Molecular conformation has been investigated through correlation between the 35Cl NQR frequencies and charge on the chlorine atoms. Its vibrational spectra has been studied. Its reaction with SO2 has been investigated. It participates in the synthesis of [(dimethylamino)phosphoryl] bis(difluoromethylene)phosphonate.

Application

Dimethylphosphoramidous dichloride (N,N-dimethylamidodichlorophosphite) may be used in the synthesis of the following:
  • N,N-dimethylamidotetrachlorophosphorane
  • O-propylchloroformimino-N,N-dimethylamidochlorophosphate
  • 1,3-bisarylsulphonyl-1,3,2,4-diazadiphosphetidines

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G K Surya Prakash et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(36), 15693-15698 (2010-08-21)
It is difficult to overestimate the importance of nucleoside triphosphates in cellular chemistry: They are the building blocks for DNA and RNA and important sources of energy. Modifications of biologically important organic molecules with fluorine are of great interest to
35Cl NQR spectra of phosphorus chlorides and their molecular conformations in crystals. Part 1. Phosphorus (III) chlorides RPCl2.
Kozlov ES, et al.
Journal of Molecular Structure, 550, 167-175 (2000)
Reaction of 1, 1-dichloro-1-nitrosobutane with N, N-dimethylamidodichlorophosphite.
Sokolov VB, et al.
Bulletin of the Academy of Sciences of the USSR, Division of chemical science, 38(6), 1296-1298 (1989)
Reactions of sulfur dioxide with aminophosphines.
Light RW and Paine RT.
Phosph. Sulfur Relat. Elem., 8(3), 255-258 (1980)
Dimethylaminodimethylphosphine1.
Burg AB and Slota Jr PJ.
Journal of the American Chemical Society, 80(5), 1107-1109 (1958)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service