Skip to Content
Merck
All Photos(1)

Key Documents

563919

Sigma-Aldrich

Aluminum chloride

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Aluminium trichloride, Aluminum(III) chloride, Trichloroaluminum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlCl3
CAS Number:
Molecular Weight:
133.34
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38120507
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 100 °C)

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: aluminum

impurities

≤15.0 ppm Trace Metal Analysis

mp

190 °C (lit.)

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum chloride is a strong Lewis acid and a major industrial catalyst. It is also used as a dopant/catalyst modifier to fabricate battery materials and solar cells.

Application

Aluminum chloride can be used:
  • To prepare cost-effective ionic liquid electrolyte for high performance aluminum ion battery.
  • To fabricate modified SnO2 triple layered photoanode for dye sensitized solar cells. This leads to suppression of electron recombination and five-fold enhancement of efficiency.
  • As a precursor to synthesize CsPbCl3 perovskite nanocrystals for bright white light-emitting diodes. It improves photoluminescence quantum yield.

Features and Benefits

  • High purity with trace metal analysis (=< 15 ppm) makes suitable for batteries.
  • High water solubility
  • Stable under battery operation conditions

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. Aluminum nitride can be synthesized from the elemnts with AlCl3 added to help facilitate crystallization.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aluminium, Gallium, Indium and Thallium
Chemistry of elements, 216-267 (1997)
Reactions of Diazirines with Aluminum Chloride: Lewis Acid-Mediated Carbene Generation and Friedel- Crafts Reactions
Moss RA, et al.
Journal of the American Chemical Society, 122(40), 9878-9879 (2000)
Reaction of xylenes with tert.-butylchloride in presence of anhydrous aluminium chloride
Ismail M, et al.
The Journal of Physical Chemistry B, 106(47), 12140-12143 (2008)
Lewis-acid catalyzed ene reactions
Snider BB
Accounts of Chemical Research, 13(11), 426-432 (1980)
Low-temperature modification of mesoporous MCM-41 material with sublimated aluminum chloride in vacuum
Xu M, et al.
The Journal of Physical Chemistry B, 106(47), 12140-12143 (2002)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service