Skip to Content
Merck
All Photos(2)

Key Documents

391522

Sigma-Aldrich

α-Methylhydrocinnamic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(CH3)CO2H
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

167-168 °C/23 mmHg (lit.)

mp

39-41 °C (lit.)

density

1.065 g/mL at 25 °C (lit.)

SMILES string

CC(Cc1ccccc1)C(O)=O

InChI

1S/C10H12O2/c1-8(10(11)12)7-9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,11,12)

InChI key

MCIIDRLDHRQKPH-UHFFFAOYSA-N

General description

α-Methylhydrocinnamic acid (2-methyl-3-phenylpropionic acid, 2-benzylpropionic acid) is a cinnamic acid derivative. Its synthesis by the asymmetric hydrogenation of α-methylcinnamic acid has been reported. α-Methylhydrocinnamic acid, a short chain fatty acid derivative (SCFAD), has been reported to correct the cystic fibrosis transmembrane conductance regulator (ΔF508-CFTR) defect. Conformational studies of 2-methyl-3-phenylpropionic acid has been investigated by NMR spectroscopy. The enantiomers of 2-benzylpropionic acid has been reported to be synthesized using a lipase-catalyzed resolution. (S) (+)-2-methyl-3-phenylpropionic acid participates in the synthesis of optically active (R)-5-methyl-6-phenylhexanoyl azide. L-2-Methyl-3.phenylpropionic acid has been reported to be an inhibitor of carboxypeptidase A. Polymer-supported “Evans” oxazolidinone mediated solid phase asymmetric has been employed in the synthesis of (S)-2-methyl-3-phenylpropionic acid.

Application

α-Methylhydrocinnamic acid is suitable for use in the comparative study to investigate the γ-globin inducibility of short-chain fatty acid derivatives (SCFADs) in mice. It may be used as a histone deacetylase (HDAC) inhibitor in the comparative study to investigate the EGFP-induction potency of a number of HDAC inhibitors. It may be used in the study to investigate the selectivity of the sensor based on imprinted poly(o-phenylenediamine) (iPoPD).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of chiral building blocks for selective adenosine receptor agents. Lipase-catalyzed resolution of 2-benzylpropanol and 2-benzylpropionic acid.
Delinck DL and Margolin AL.
Tetrahedron Letters, 31(47), 6797-6798 (1990)
Jörgen Samuelsson et al.
Journal of chromatography. A, 1163(1-2), 177-189 (2007-07-07)
A systematic study was made to explain the large improvements in separation performance and capacity of basic compounds at alkaline conditions. The adsorption of three probe components was investigated on four alkaline-stable silica-based C18 columns at three different pH-levels: 3
H Peng et al.
The Analyst, 126(2), 189-194 (2001-03-10)
The preparation and characterization of electrosynthesized poly(o-phenylenediamine) (iPoPD) as a molecular imprinting material were studied by an in situ quartz crystal impedance method. The changes of delta f0, delta R1, delta L1 and delta C0 suggest that the polymer film
The preparation and first application of a polymer-supported ?Evans? oxazolidinone.
Allin SM and Shuttleworth SJ.
Tetrahedron Letters, 37(44), 8023-8026 (1996)
Yishan Liu et al.
Journal of hazardous materials, 192(3), 1633-1640 (2011-07-29)
Microbial degradation of the chiral 2-phenylbutyric acid (2-PBA), a metabolite of surfactant linear alkylbenzene sulfonates (LAS), was investigated using both racemic and enantiomer-pure compounds together with quantitative stereoselective analyses. A pure culture of bacteria, identified as Xanthobacter flavus strain PA1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service