Skip to Content
Merck

116017

2-Carboxybenzaldehyde

97%

Synonym(s):

Phthalaldehydic acid, 2-Formylbenzoic acid, 3-Hydroxyphthalide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
204-342-3
Beilstein/REAXYS Number:
742381
MDL number:
Assay:
97%
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

form

solid

mp

94-96 °C (lit.)

functional group

aldehyde, carboxylic acid

SMILES string

OC(=O)c1ccccc1C=O

InChI

1S/C8H6O3/c9-5-6-3-1-2-4-7(6)8(10)11/h1-5H,(H,10,11)

InChI key

DYNFCHNNOHNJFG-UHFFFAOYSA-N

General description

2-Carboxybenzaldehyde is an aldehydic acid that can be used to synthesize functionalized diazaphospholanes.

Application

2-Carboxybenzaldehyde is a metabolite of the biodegradation of luoranthene using two pure bacterial strains, Pasteurella sp. IFA (B-2) and Mycobacterium sp. PYR-1 (AM). It has been used to synthesize a series of N-substituted isoindolinones by reductive C-N coupling and intramolecular amidation with amines.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library