Skip to Content
Merck
All Photos(2)

Key Documents

158518

Sigma-Aldrich

Phenoxyacetic acid

98%

Synonym(s):

Glycolic acid phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
907949
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

solid

mp

98-100 °C (lit.)

solubility

ethanol: soluble 10%, clear, colorless

SMILES string

OC(=O)COc1ccccc1

InChI

1S/C8H8O3/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

LCPDWSOZIOUXRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenoxyacetic acid is a herbicide[1] and its photodegradation using TiO2 as photocatalyst has been studied.[2] Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photocatalyzed Degradation of Phenol, 2, 4-Dichlorophenol, Phenoxyacetic Acid and 2, 4-Dichlorophenoxyacetic Acid over SupportedTiO2 in a Flow System.
Trillas M, et al.
Journal of Chemical Technology and Biotechnology, 67(3), 237-242 (1996)
Selective separation of penicillin V from phenoxyacetic acid using liquid membranes.
Cascaval D, et al.
Biochemical Engineering Journal, 55(1), 45-50 (2000)
Rudina Bleta et al.
Langmuir : the ACS journal of surfaces and colloids, 30(39), 11812-11822 (2014-09-16)
A series of mesoporous titania photocatalysts with tailorable structural and textural characteristics was prepared in aqueous phase via a colloidal self-assembly approach using various cyclodextrins (CDs) as structure-directing agents. The photocatalysts and the structure-directing agents were characterized at different stages
Ming-Kung Yeh et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 27(1), 49-54 (2011-01-15)
Dynamic liquid-liquid-liquid microextraction coupled with ion-pair liquid chromatography (IP-LC) and photodiode array detection was developed and used for the extraction and analysis of chlorinated phenoxyacetic acids (CPAs) and chlorophenols (CPs) from water samples. An organic extraction solvent mixture was chosen
Giuseppe Fracchiolla et al.
ChemMedChem, 2(5), 641-654 (2007-03-16)
Peroxisome proliferator-activated receptors (PPARs) are ligand-activated transcription factors that govern lipid and glucose homeostasis, and play a central role in cardiovascular disease, obesity, and diabetes. Thus, there is significant interest in developing new and specific agonists for these receptors. Herein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service