Skip to Content
Merck
All Photos(3)

Key Documents

339067

Sigma-Aldrich

Boric acid

99.97% trace metals basis

Synonym(s):

Boracic acid, Borofax

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38120104
PubChem Substance ID:
NACRES:
NA.23

Agency

suitable for SM 5210

vapor pressure

2.6 mmHg ( 20 °C)

Assay

99.97% trace metals basis

form

powder or crystals

mp

160 °C (dec.) (lit.)

solubility

water: soluble

density

1.440 g/cm3

application(s)

histology

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Boric acid is a weak monobasic acid, it accepts OH- ions, hence is a Lewis acid. In boric acid, B is sp2 hybridized, forming a planar triangle structure. The principal oxide of boron, B2O3, is obtained as a vitreous solid by dehydration of boric acid at red heat.

Application

Boric acid was used as a constituent in mineral sacrificial paste. 1 It has been used as a buffer in electrodeposition of nickel. 2

Packaging

Packaged in poly bottles

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
American journal of physiology. Cell physiology, 306(10), C943-C960 (2014-03-22)
The human solute carrier (SLC26) family of anion transporters consists of 10 members (SLCA1-11, SLCA10 being a pseudogene) that encode membrane proteins containing ~12 transmembrane (TM) segments with putative N-glycosylation sites (-NXS/T-) in extracellular loops and a COOH-terminal cytosolic STAS
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic
Hye Ryeo Lee et al.
Journal of chromatography. A, 1346, 117-122 (2014-05-09)
In liquid phase microextraction, high enrichment factors can be obtained using an acceptor phase of small volume. By hanging an acceptor drop at the separation capillary tip, single drop microextraction (SDME) can be in-line coupled with capillary electrophoresis (CE). The
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed
Sunita A Ramesh et al.
Nature communications, 6, 7879-7879 (2015-07-30)
The non-protein amino acid, gamma-aminobutyric acid (GABA) rapidly accumulates in plant tissues in response to biotic and abiotic stress, and regulates plant growth. Until now it was not known whether GABA exerts its effects in plants through the regulation of

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service