Skip to Content
Merck
All Photos(1)

Documents

33522

Sigma-Aldrich

Phenol-1,2-dichlorobenzene mixture

1:1, 49.5-50.5% phenol basis (GC), 49.5-50.5% 1,2-dichlorbenzene basis (GC)

Synonym(s):

o-Dichlorobenzene/Phenol (mixture 1:1)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

form

liquid

composition

1,2-dichlorbenzene, 49.5-50.5% GC
phenol, 49.5-50.5% GC

SMILES string

Oc1ccccc1.Clc2ccccc2Cl

InChI

1S/C6H4Cl2.C6H6O/c7-5-3-1-2-4-6(5)8;7-6-4-2-1-3-5-6/h1-4H;1-5,7H

InChI key

WEDPGDLPFLPTFH-UHFFFAOYSA-N

General description

Phenol-1,2-dichlorobenzene mixture is a mixture of phenol and 1,2-dichlorobenzene in the ratio of 1:1. 1,2-Dichlorobenzene/Phenol mixture (50/50wt%) has been used as solvent for poly(butylene succinate) (PBS) during the evaluation of intrinsic viscosity of PBS by Huggins and Kramer plots, and from the single-point methods.

Application

Phenol-1,2-dichlorobenzene mixture may be used as a solvent to measure the intrinsic viscosity of a polymer sample using an Ubbelohde viscometer.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system,Kidney,Liver,Skin, Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of processing conditions on the thermal and electrical conductivity of poly (butylene terephthalate) nanocomposites prepared via ring-opening polymerization.
Colonna S, et al.
Materials & Design, 119, 124-132 (2017)
The effects of long chain branching of polypropylene and chain extension of poly (ethylene terephthalate) on the thermal behavior, rheology and morphology of their blends.
Jahani Y, et al.
Royal Society of Chemistry Advances, 5(28), 21620-21628 (2015)
Solution viscosity-molar mass relationships for poly (butylene succinate) and discussion on molar mass analysis.
Charlier Q, et al.
Express Polymer Letters, 9(5), 424-434 (2015)
Molecular modelling of polyphthalamides thermal properties: Comparison between modelling and experimental results.
Cousin T, et al.
Polymer, 53(15), 3203-3210 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service