Skip to Content
Merck
All Photos(1)

Key Documents

332615

Sigma-Aldrich

Methyl-β-cyclodextrin

average Mn 1310

Synonym(s):

MβCD, MEBCD, Methyl-β-CD, Methyl-beta-cyclodextrin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

form

powder

optical activity

[α]25/D +159°, c = 1 in H2O

mol wt

average Mn 1310

extent of labeling

1.6-2.0 mol CH3 per unit anhydroglucose

color

white

mp

180-182 °C (lit.)

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C(OC8OC(C1C(C8OC)O)COC)C(C7OC)O)COC)C(C6OC)O)COC)C(C5OC)O)COC)C(C4OC)O)COC)C(C3OC)O)COC)C(C2OC)O)COC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3

InChI key

QGKBSGBYSPTPKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl-β-cyclodextrin (MβCD) is a macrocyclic compound that can form inclusion complexes with a number of guest molecules. It shows higher solubility in aqueous solutions and greater solubilizing and complexing power compared to the parent β-cyclodextrin. MβCD can be used to enhance the enzyme activity and enantioselectivity of subtilizing enzyme in organic solvents.

Application

Methyl-β-cyclodextrin is a cholesterol-removing agent mainly used for lipid raft disruption. It may also be used for detergent removal from protein-lipid-detergent ternary mixture to produce 2D membrane protein crystals due to its high solubility and affinity for detergents generally used in membrane protein chemistry.
Methyl-β-cyclodextrin may be used as:
  • A buffer additive for the effective separation of aminophospholipids by modified micellar electrokinetic capillary chromatography (MEKC) coupled with laser-induced fluorescence detection.
  • A mobile phase additive for the analysis of R,S-naproxen mixtures in commercial tablet formulations by conventional and nano-liquid chromatography (nano-LC).
  • An analytical standard for the determination of the analyte in biological samples by high performance liquid chromatography (HPLC).
  • A chiral selector for the resolution of repaglinide enantiomers in pharmaceutical formulations by capillary electrophoresis.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

368.6 °F

Flash Point(C)

187 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tea polyphenol epigallocatechin?3?gallate associates with plasma membrane lipid rafts: Lipid rafts mediate anti?allergic action of the catechin.
Tachibana H, et al.
Biofactors, 21(1?4), 383-385 (2004)
P Y Grosse et al.
Journal of chromatography. B, Biomedical sciences and applications, 694(1), 219-226 (1997-06-20)
This paper describes a high-performance liquid chromatographic method with fluorescence detection for the analysis of methyl-beta-cyclodextrin (MEBCD) in plasma and cell lysate, after in situ complexation with 1-naphthol. The size-exclusion HPLC column packed with TSK 3000 SW gel, was equilibrated
Controlled 2D crystallization of membrane proteins using methyl-?-cyclodextrin.
Signorell GA, et al.
Journal of Structural Biology, 157(2), 321-328 (2007)
Andreza Ribeiro et al.
AAPS PharmSciTech, 9(4), 1102-1109 (2008-11-01)
The aim of this study is to confirm the formation of inclusion complexes between miconazole (MCZ) and two derivatives of beta-cyclodextrin, methyl-beta-cyclodextrin (MbetaCD) and 2-hydroxypropyl-beta-cyclodextrin (HPbetaCD) in aqueous solution by phase solubility studies. Inclusion complexes with MbetaCD in the solid
Depletion of membrane cholesterol causes ligand-independent activation of Fas and apoptosis.
Gniadecki R.
Biochemical and Biophysical Research Communications, 320(1), 165-169 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service