Skip to Content

M7439

Methyl-β-cyclodextrin

Synonym(s):

Methyl-beta-cyclodextrin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

impurities

<10.0% water (Karl Fischer)

color

white

mp

180-182 °C (lit.)

application(s)

advanced drug delivery

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C(OC8OC(C1C(C8OC)O)COC)C(C7OC)O)COC)C(C6OC)O)COC)C(C5OC)O)COC)C(C4OC)O)COC)C(C3OC)O)COC)C(C2OC)O)COC

InChI

1S/C56H98O35/c1-64-15-22-36-29(57)43(71-8)50(78-22)86-37-23(16-65-2)80-52(45(73-10)30(37)58)88-39-25(18-67-4)82-54(47(75-12)32(39)60)90-41-27(20-69-6)84-56(49(77-14)34(41)62)91-42-28(21-70-7)83-55(48(76-13)35(42)63)89-40-26(19-68-5)81-53(46(74-11)33(40)61)87-38-24(17-66-3)79-51(85-36)44(72-9)31(38)59/h22-63H,15-21H2,1-14H3

InChI key

QGKBSGBYSPTPKJ-UHFFFAOYSA-N

General description

Methyl-β-cyclodextrin is a small cyclic heptasaccharide.

Application

Methyl-β-cyclodextrin has been used as a cholesterol chelator:
  • to study its effects on mice cardiomyocytes
  • to study its effects on human corneal stromal fibroblasts
  • to study its effects on the junctophilin-2 (JPH2) pool

Biochem/physiol Actions

Methyl-β-cyclodextrin participates in increasing the permeability of cells. It also plays a role in depolymerizing the actin cytoskeleton in cells. Methyl-β-cyclodextrin is involved in attracting cholesterol from the plasma. It participates in activating extracellular receptor kinase in certain cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Methyl-β-cyclodextrin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

368.6 °F

flash_point_c

187 °C

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library