Skip to Content
Merck
All Photos(2)

Key Documents

190357

Sigma-Aldrich

(R)-(−)-2-Phenylglycinol

98%, for peptide synthesis

Synonym(s):

(R)-2-Amino-2-phenylethanol, D-(−)-α-Phenylglycinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(NH2)CH2OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
2935848
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

(R)-(−)-2-Phenylglycinol, 98%

Assay

98%

optical activity

[α]24/D −31.7°, c = 0.76 in 1 M HCl

optical purity

ee: 99% (GLC)

reaction suitability

reaction type: solution phase peptide synthesis

mp

75-77 °C (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](CO)c1ccccc1

InChI

1S/C8H11NO/c9-8(6-10)7-4-2-1-3-5-7/h1-5,8,10H,6,9H2/t8-/m0/s1

InChI key

IJXJGQCXFSSHNL-QMMMGPOBSA-N

Application

Amino alcohol used to prepare a chiral imine or oxazolidine from ethyl trifluoropyruvate. These intermediates were then employed in a synthesis of both enantiomers of α-trifluoromethylproline.
Chiral β−amino alcohol used as a synthetic building block.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Grégory Chaume et al.
Organic letters, 8(26), 6123-6126 (2006-12-15)
[Structure: see text] A concise synthesis of both enantiomers of alpha-Tfm-proline and (S)-alpha-Tfm-prolinol from ethyl trifluoropyruvate is reported. The key step is a diastereoselective allylation reaction of ethyl trifluoropyruvate and (R)-phenylglycinol-based oxazolidines or imine. The lactone obtained by cyclization of
Tetrahedron Letters, 45, 5287-5287 (2004)
Amedjkouh, M.; Westerlund, K.
Tetrahedron Letters, 45, 5175-5175 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service