Skip to Content
Merck
All Photos(3)

Key Documents

325171

Sigma-Aldrich

Silicon nitride

predominantly α-phase, ≤10 micron

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si3N4
CAS Number:
Molecular Weight:
140.28
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

particle size

≤10 micron

density

3.44 g/mL at 25 °C (lit.)

SMILES string

N12[Si]34N5[Si]16N3[Si]25N46

InChI

1S/N4Si3/c1-5-2-6(1)3(5)7(1,2)4(5)6

InChI key

HQVNEWCFYHHQES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Mazzotta et al.
Nanotechnology, 23(41), 415304-415304 (2012-09-29)
One of the primary advantages of nanoscale sensors is that they often can provide conceptually new ways of performing sensing that are not feasible with their large-scale analogs. For example, the small size of nanoscale sensor elements, such as plasmonic
Madeline J Dukes et al.
Chemical communications (Cambridge, England), 49(29), 3007-3009 (2013-03-06)
Gold nanorods are widely known for their photothermal properties to treat solid tumors. Our work demonstrates the unrealized capacity to image these reagents in liquid at high resolution using Transmission Electron Microscopy (TEM). Here we perform the first atomic measurements
Jinglin Kong et al.
Journal of nanoscience and nanotechnology, 13(6), 4010-4016 (2013-07-19)
In recent years, nanopore has attracted broad attention as an essential technique for DNA sequencing and nanoparticle sensing. This work presented a fundamental study of nanoparticle translocation through silicon nitride nanopores. Since particles with rather high charge to mass ratio
Christophe Baker et al.
Optics express, 20(27), 29076-29089 (2012-12-25)
We report time domain observations of optical instability in high Q silicon nitride whispering gallery disk resonators. At low laser power the transmitted optical power through the disk looks chaotic. At higher power, the optical output settles into a stable
Benjamin N Miles et al.
Chemical Society reviews, 42(1), 15-28 (2012-09-20)
This tutorial review will introduce and explore the fundamental aspects of nanopore (bio)sensing, fabrication, modification, and the emerging technologies and applications that both intrigue and inspire those working in and around the field. Although nanopores can be classified into two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service