Skip to Content
Merck
All Photos(3)

Key Documents

378097

Sigma-Aldrich

Silicon carbide

-200 mesh particle size

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiC
CAS Number:
Molecular Weight:
40.10
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

particle size

-200 mesh

mp

2700 °C (lit.)

density

3.22 g/mL at 25 °C (lit.)

SMILES string

[C-]#[Si+]

InChI

1S/CSi/c1-2

InChI key

HBMJWWWQQXIZIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silicon carbide (SiC) is a semiconductor based material that consists of a closed packed stacking of double layers of silicon and carbon. It has an excellent thermo-mechanical property that makes it useful in a variety of electronic and optoelectronic applications.

Application

SiC is majorly used as a base material for applications such as micro-structures, opto-electronic devices(OLEDs), UV detectors, high temperature electronics (nuclear electronics), high frequency devices

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Properties of silicon carbide (1995)
A high-density, high-efficiency, isolated on-board vehicle battery charger utilizing silicon carbide power devices
Whitaker B, et al.
IEEE Transactions on Power Electronics, 29(5), 2606-2617 (2013)
Optical polarization of nuclear spins in silicon carbide
Falk AL, et al.
Physical Review Letters, 114(24), 247603-247603 (2015)
Development of SiC-Si composites with fine-grained SiC microstructures
Wilhelm M, et al.
J. Eur. Ceram. Soc., 19(12), 2155-2163 (1999)
Fundamentals of silicon carbide technology: growth, characterization, devices and applications (2014)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service