Skip to Content
Merck
N-Phenyl-bis(trifluoromethanesulfonimide) ≥98.0% (HPLC)

N-Phenyl-bis(trifluoromethanesulfonimide)

N-Phenyl-bis(trifluoromethanesulfonimide) ≥98.0% (HPLC)
Synonym(s):
1,1,1-Trifluoro-N-phenyl-N-[(trifluoromethyl)sulfonyl]methanesulfonamide, N,N-Bis(trifluoromethylsulfonyl)aniline, N-Phenyl-trifluoromethanesulfonimide, NSC 240874, Phenyl triflimide
Linear Formula:
C6H5N(SO2CF3)2
CAS Number:
Molecular Weight:
357.25

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2