Skip to Content
Merck
All Photos(1)

Key Documents

291013

Sigma-Aldrich

Chlorotriisobutylsilane

97%

Synonym(s):

Triisobutylchlorosilane, Triisobutylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]3SiCl
CAS Number:
Molecular Weight:
234.88
Beilstein:
2409275
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.448 (lit.)

density

0.875 g/mL at 25 °C (lit.)

SMILES string

CC(C)C[Si](Cl)(CC(C)C)CC(C)C

InChI

1S/C12H27ClSi/c1-10(2)7-14(13,8-11(3)4)9-12(5)6/h10-12H,7-9H2,1-6H3

InChI key

TZPZCTBZJKZFGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorotriisobutylsilane can be used as an intermediate in the synthesis of:
  • Silicon (IV) phthalocyanines, which are used as photosensitizers for the photodynamic inactivation of bacteria.
  • Poly(4-alkylthiazole) polymers exhibiting optical and electronic properties.
  • Bis(trialkylsilyl oxide) based silicon phthalocyanines applicable as ternary additives in organic photovoltaics.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Octacationic and axially di-substituted silicon (IV) phthalocyanines for photodynamic inactivation of bacteria
van de Winckel E, et al.
Dyes and Pigments, 145, 239-245 (2017)
Bis (trialkylsilyl oxide) Silicon Phthalocyanines: Understanding the Role of Solubility in Device Performance as Ternary Additives in Organic Photovoltaics
Vebber MC, et al.
Langmuir, 36(10), 2612-2621 (2020)
Toward n-type analogues to poly (3-alkylthiophene) s: influence of side-chain variation on bulk-morphology and electron transport characteristics of head-to-tail regioregular poly (4-alkylthiazole) s
Jager J, et al.
Journal of Material Chemistry C, 4(13), 2587-2597 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service