Skip to Content
Merck
All Photos(1)

Key Documents

383767

Sigma-Aldrich

Tri(2-furyl)phosphine

Tri(2-furyl)phosphine

99%

Synonym(s):

TFP, Tri-2-furanylphosphine, Tris(2-furanyl)phosphine, Tris(o-furyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9O3P
CAS Number:
Molecular Weight:
232.17
Beilstein:
1577564
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

solid

reaction suitability

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Cycloisomerizations

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Oxidations

reagent type: ligand
reaction type: Sonogashira Coupling

reagent type: ligand
reaction type: Stille Coupling

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

bp

136 °C/4 mmHg (lit.)

mp

59-64 °C (lit.)

functional group

phosphine

SMILES string

c1coc(c1)P(c2ccco2)c3ccco3

InChI

1S/C12H9O3P/c1-4-10(13-7-1)16(11-5-2-8-14-11)12-6-3-9-15-12/h1-9H

InChI key

DLQYXUGCCKQSRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajime Monzen et al.
Journal of applied clinical medical physics, 18(5), 325-329 (2017-06-29)
Tungsten functional paper (TFP) is a novel paper-based radiation-shielding material. We measured the shielding ability of TFP against x-rays and gamma rays. The TFP was supplied in 0.3-mm-thick sheets that contained 80% tungsten powder and 20% cellulose (C
Liang-da Chiu et al.
Scientific reports, 7, 43569-43569 (2017-03-09)
Our current understanding of molecular biology provides a clear picture of how the genome, transcriptome and proteome regulate each other, but how the chemical environment of the cell plays a role in cellular regulation remains much to be studied. Here
Tim Buchholz et al.
Animals : an open access journal from MDPI, 10(9) (2020-09-23)
When using animals in biomedical research, investigators have the responsibility to ensure adequate analgesia. Currently, transdermal fentanyl patches (TFP) are often used to provide postoperative analgesia in large laboratory animals. The aim of this study was to compare the fentanyl
Jingwen Jiang et al.
DNA and cell biology, 36(10), 813-821 (2017-09-07)
Schizophrenic patients tend to have reduced incidence of some cancers due to the treatment of antipsychotic drugs with antitumor effects, such as chlorpromazine and trifluoperazine (TFP). Forkhead Box O1 (FOXO1) as tumor suppressor in many malignancies is often inactivated by
Na Zhang et al.
International journal of environmental research and public health, 17(10) (2020-05-21)
Green development is an important way to meet the challenges of ecological and environmental protection and economic growth, as well as an inevitable choice to realize China's sustainable development in the new era. The Chinese economic system is such that

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service