Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Raney
®
-Nickel
Synonym(s):
Nickel sponge
Linear Formula:
AlNi
CAS Number:
7440-02-0
Products
Brand
Sigma-Aldrich (4)
Manufacturer
pH Value
Physical Form
Chemical Composition
Particle Size (µm)
Reagent Type
Core
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
221678
Raney
®
-Nickel,
W.R. Grace and Co. Raney
®
2800, slurry, in H
2
O, active catalyst
Expand
View Pricing
510033
Raney
®
-Nickel,
W.R. Grace and Co. Raney
®
2400, slurry, in H
2
O, active catalyst
Expand
View Pricing
510041
Raney
®
-Nickel,
W.R. Grace and Co. Raney
®
4200, slurry, in H
2
O, active catalyst
Expand
View Pricing
510068
Raney
®
-Nickel,
W.R. Grace and Co. Raney
®
3202, slurry, in H
2
O, active catalyst
Expand
View Pricing