Skip to Content
Merck
All Photos(1)

Key Documents

214671

Sigma-Aldrich

3-Acetylcoumarin

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

form

solid

mp

119-122 °C (lit.)

functional group

ester
ketone

SMILES string

CC(=O)C1=Cc2ccccc2OC1=O

InChI

1S/C11H8O3/c1-7(12)9-6-8-4-2-3-5-10(8)14-11(9)13/h2-6H,1H3

InChI key

CSPIFKKOBWYOEX-UHFFFAOYSA-N

General description

FTIR and FT-Raman spectra of 3-acetylcoumarin has been reported[1]. 3-Acetylcoumarin undergoes condensation with aryl aldehydes in chloroform in the presence of piperidine to yield coumarin derivatives containing 4-arylbut-3-en-2-one moiety[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdullah Sulaiman Al-Ayed
Molecules (Basel, Switzerland), 16(12), 10292-10302 (2011-12-14)
A series of new coumarin derivatives 4 containing a 4-arylbut-3-en-2-one moiety were synthesized by condensation of 3-acetylcoumarin 1 with aryl aldehydes 2 in chloroform in the presence of piperidine. The interactions of 3-formyl-4-chlorocoumarin (3) with nitrogen-containg nucleophiles leading to the
X L Huang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 31(6), 431-436 (1996-01-01)
Twenty-five 3-acetylcoumarin derivatives were synthesized among which twenty-two were not reported before. Antimutagenic activity screen in vitro has shown that some of these compounds have various activities. The structure and activity relationship for 5-, 7-, 8-substituents has been studied. Pharmacological
Wafaa S Hamama et al.
Archiv der Pharmazie, 344(11), 710-718 (2011-09-29)
3-Acetylcoumarin (1) was utilized as a key intermediate for the synthesis of 2-aminothiazole derivative 3 via bromination of 1 to afford acetylbromide 2 followed by treatment with thiourea or via Biginelli reaction of 1. Treatment of 3 with 5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 2-methyl-4H-benzo[d][1,3]oxazin-4-one
Jhi Biau Foo et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 31(4), 505-515 (2018-04-07)
Copper complexes have been widely studied for the anti-tumour application as cancer cells are reported to take up greater amounts of copper than normal cells. Preliminary study revealed that the newly synthesised copper complex [Cu(SBCM)2] displayed marked anti-proliferative towards triple-negative
V Arjunan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 109, 79-89 (2013-03-19)
3-Acetylcoumarin (3AC) was synthesised by a Knoevenagel reaction. Conformational analysis using the B3LYP method was also carried out to determine the most stable conformation of the compound. FTIR and FT-Raman spectra of 3AC have been recorded in the range 4000-400

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service