Skip to Content
Merck
All Photos(1)

Key Documents

C85603

Sigma-Aldrich

Coumarin-3-carboxylic acid

99%

Synonym(s):

2-Oxo-2H-1-benzopyran-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6O4
CAS Number:
Molecular Weight:
190.15
Beilstein:
154276
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

powder

mp

189-192 °C (lit.)

SMILES string

OC(=O)C1=Cc2ccccc2OC1=O

InChI

1S/C10H6O4/c11-9(12)7-5-6-3-1-2-4-8(6)14-10(7)13/h1-5H,(H,11,12)

InChI key

ACMLKANOGIVEPB-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Ali et al.
Free radical research, 32(5), 429-438 (2000-04-15)
The effect of lactic acid (lactate) on Fenton based hydroxyl radical (*OH) production was studied by spin trapping, ESR, and fluorescence methods using DMPO and coumarin-3-carboxylic acid (3-CCA) as the *OH traps respectively. The *OH adduct formation was inhibited by
Rajsekhar Roy et al.
ACS omega, 5(30), 18628-18641 (2020-08-11)
In Alzheimer's disease (AD), insoluble Aβ42 peptide fragments self-aggregate and form oligomers and fibrils in the brain, causing neurotoxicity. Further, the presence of redox-active metal ions such as Cu2+ enhances the aggregation process through chelation with these Aβ42 aggregates as
Bhumika Thati et al.
Cancer letters, 248(2), 321-331 (2006-09-26)
The chemotherapeutic potential of coumarin-3-carboxylic acid (C-3-COOH) and a series of three hydroxylated coumarin-3-carboxylic acid ligands, namely 6-hydroxy-coumarin-3-carboxylic acid (6-OH-C-3-COOH), 7-hydroxy-coumarin-3-carboxylic acid (7-OH-C-3-COOH) and 8-hydroxy-coumarin-3-carboxylic acid (8-OH-C-3-COOH), along with their corresponding silver-based complexes, namely 6-hydroxycoumarin-3-carboxylatosilver (6-OH-C-COO-Ag), 7-hydroxycoumarin-3-carboxylatosilver (7-OH-C-COO-Ag) and 8-hydroxycoumarin-3-carboxylatosilver
Grigory V Andrievsky et al.
Free radical biology & medicine, 47(6), 786-793 (2009-06-23)
Aqueous solutions of highly stable supramolecular donor-acceptor complexes of chemically nonmodified pristine C(60) fullerene molecules with H(2)O molecules (hydrated C(60) fullerene-C(60)HyFn) and their labile nano-sized clusters were examined for their antioxidant effects on removal of hydroxyl radicals (.OH) and protecting
Judith A K Howard et al.
Acta crystallographica. Section B, Structural science, 65(Pt 2), 230-237 (2009-03-21)
As part of an ongoing series of experimental charge-density investigations into the intra- and intermolecular interactions present in compounds which undergo solid-state [2 + 2] cycloaddition reactions, the charge-density analyses of trans-cinnamic acid and coumarin-3-carboxylic acid are reported. Thus, high-resolution

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service